Skip to Content
Merck
All Photos(1)

Key Documents

R7385

Sigma-Aldrich

Phosphoramidon disodium salt

powder, ≥97% (HPLC)

Synonym(s):

N-(α-Rhamno­pyranosyl­oxy­hydroxy­phosphinyl)-Leu-Trp disodium salt, N-(α-Rhamno­pyranosyl­phos­phono)-L-leucyl-L-tryptophan disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32N3Na2O10P
CAS Number:
Molecular Weight:
587.47
Beilstein:
8105669
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Product Name

Phosphoramidon disodium salt, ≥97% (HPLC)

biological source

Streptomyces sp.

Assay

≥97% (HPLC)

form

powder

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

[Na+].[Na+].CC(C)C[C@H](NP([O-])(=O)O[C@@H]1O[C@@H](C)[C@H](O)[C@@H](O)[C@H]1O)C(=O)N[C@@H](Cc2c[nH]c3ccccc23)C([O-])=O

InChI

1S/C23H34N3O10P.2Na/c1-11(2)8-16(26-37(33,34)36-23-20(29)19(28)18(27)12(3)35-23)21(30)25-17(22(31)32)9-13-10-24-15-7-5-4-6-14(13)15;;/h4-7,10-12,16-20,23-24,27-29H,8-9H2,1-3H3,(H,25,30)(H,31,32)(H2,26,33,34);;/q;2*+1/p-2/t12-,16-,17-,18-,19+,20+,23-;;/m0../s1

InChI key

OQKHVXFOYFBMDJ-ODIUWQMJSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phosphoramidon [N-(α-rhamnopyranosyl-oxyhydroxyphosphinyl)-Leu-Trp] is a competitive inhibitor of several soluble zinc metalloproteases. Phosphoramidate is produced naturally by Streptomyces tanashiensis.

Application

Identification of phosphoramidon as a metallo-endopeptidase inhibitor was used to characterize endothelin converting enzyme as a metallo-endopeptidase. Phosphoramidon increases the production of β-amyloid protein, but not amyloid precursor protein, indicating that abnormal amyloid processing in Alzheimer′s disease may be carried out by a metallo-endopeptidase.
Phosphoramidon disodium salt has been used:
  • as the neprilysin (NEP) inhibitor: in amyloid beta (Aβ) uptake and degradation
  • to study its effect on keratinase activity
  • to treat primary microglia from wild-type mice

Biochem/physiol Actions

Phosphoramidon can inhibit ZMPSTE24, an integral membrane protein zinc metalloprotease. It is capable of blocking thermolysin.
Phosphoramidon is a potent inhibitor of thermolysin and other metallo-endopeptidases. It trongly inhibits mammalian enkephalinase and it does not inhibit trypsin, papain, chymotrypsin or pepsin. It weakly inhibits collagenase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermolysin and Related Bacillus Metallopeptidases
Handbook of Proteolytic Enzymes, 1, 540-553 (2013)
J R Wu-Wong et al.
Molecular pharmacology, 44(2), 422-429 (1993-08-01)
Endothelin (ET) is generated from prepro-ET by dibasic pair proteolysis, followed by specific proteolytic cleavage between Trp21 and Val22. Currently, intense research efforts are focused on the investigation of a metalloprotease-like ET-converting enzyme that is inhibited by phosphoramidon but not
Deacetylation of TFEB promotes fibrillar A beta degradation by upregulating lysosomal biogenesis in microglia
Bao J, et al.
Protein & cell, 7(6), 417-433 (2016)
SIRT1 facilitates amyloid beta peptide degradation by upregulating lysosome number in primary astrocytes
Li MZ, et al.
Neural Regeneration Research, 13(11), 2005-2005 (2018)
Phosphoramidon inhibits the integral membrane protein zinc metalloprotease ZMPSTE24
Goblirsch BR, et al.
Acta crystallographica. Section D, Structural biology, 74(Pt 8), 739-739 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service