Skip to Content
Merck
All Photos(2)

Key Documents

D4292

Sigma-Aldrich

3,3′-Dioctadecyloxacarbocyanine perchlorate

≥98% purity (TLC), powder

Synonym(s):

DiOC18(3), ‘DiO’

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H85ClN2O6
CAS Number:
Molecular Weight:
881.70
Beilstein:
4122771
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

3,3′-Dioctadecyloxacarbocyanine perchlorate,

Assay

≥98% (TLC)

form

powder

technique(s)

titration: suitable

color

orange

solubility

chloroform: 50 mg/mL

fluorescence

λex 484 nm; λem 501 nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[O-]Cl(=O)(=O)=O.CCCCCCCCCCCCCCCCCCN1C(\Oc2ccccc12)=C/C=C/c3oc4ccccc4[n+]3CCCCCCCCCCCCCCCCCC

InChI

1S/C53H85N2O2.ClHO4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-37-46-54-48-40-33-35-42-50(48)56-52(54)44-39-45-53-55(49-41-34-36-43-51(49)57-53)47-38-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;2-1(3,4)5/h33-36,39-45H,3-32,37-38,46-47H2,1-2H3;(H,2,3,4,5)/q+1;/p-1

InChI key

GFZPJHFJZGRWMQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

3,3′-Dioctadecyloxacarbocyanine perchlorate has been used for lipid bilayer labeling in antigen-presenting cells. It has also been used as a lipophilic dye for labeling in zebrafish embryos.

Biochem/physiol Actions

3,3′-Dioctadecyloxacarbocyanine perchlorate is a fluorescent probe of lipid bilayer model membranes. It is a fluorescent marker for living cells in culture.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hassan Nehme et al.
PloS one, 13(1), e0189950-e0189950 (2018-01-04)
Bacterial antibiotic resistance is an emerging public health problem worldwide; therefore, new therapeutic strategies are needed. Many studies have described antipsychotic compounds that present antibacterial activity. Hence, the aims of this study were to evaluate the in vitro antibacterial activity
Dynamic behavior of fluorescent probes in lipid bilayer model membranes.
R A Badley et al.
Biochemistry, 12(2), 268-275 (1973-01-16)
Jayesh A Kulkarni et al.
Nanoscale, 11(18), 9023-9031 (2019-04-26)
The success of Onpattro™ (patisiran) clearly demonstrates the utility of lipid nanoparticle (LNP) systems for enabling gene therapies. These systems are composed of ionizable cationic lipids, phospholipid, cholesterol, and polyethylene glycol (PEG)-lipids, and are produced through rapid-mixing of an ethanolic-lipid
Kunxi Zhang et al.
Acta biomaterialia, 51, 246-257 (2017-01-18)
Vascularization is of great importance to adipose tissue regeneration. Here we introduced a paradigm that using scaffold to induce ASC spheroids, so to promote vascularized adipose tissue regeneration. Poly (l-glutamic acid) (PLGA) was activated by EDC, followed by being cross-linked
Xiaochun Hou et al.
SLAS technology, 22(4), 447-453 (2016-06-22)
Nanoparticles are emerging transdermal delivery systems. Their size and surface properties determine their efficacy and efficiency to penetrate through the skin layers. This work utilizes three-dimensional (3D) bioprinting technology to generate a simplified artificial skin model to rapidly screen nanoparticles

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service