Skip to Content
Merck
All Photos(1)

Documents

42364

Sigma-Aldrich

1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate

BioReagent, suitable for fluorescence, ≥98.0% (TLC)

Synonym(s):

DiI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H97ClN2O4
CAS Number:
Molecular Weight:
933.87
Beilstein:
8251870
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥98.0% (TLC)

mp

68 °C (dec.) (lit.)

solubility

DMF: soluble
DMSO: soluble
methanol: soluble

fluorescence

λex 550 nm; λem 567 nm in phosphate buffer/SDS pH 7.0

suitability

suitable for fluorescence

SMILES string

[O-]Cl(=O)(=O)=O.CCCCCCCCCCCCCCCCCCN1c2ccccc2C(C)(C)/C1=C\C=C\C3=[N+](CCCCCCCCCCCCCCCCCC)c4ccccc4C3(C)C

InChI

1S/C59H97N2.ClHO4/c1-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-41-50-60-54-46-39-37-44-52(54)58(3,4)56(60)48-43-49-57-59(5,6)53-45-38-40-47-55(53)61(57)51-42-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-2;2-1(3,4)5/h37-40,43-49H,7-36,41-42,50-51H2,1-6H3;(H,2,3,4,5)/q+1;/p-1

InChI key

JVXZRNYCRFIEGV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

DiI is a Dye Aye, a chemical with CAS number 41085-99-8. It is a fluorescent lipophilic cationic indocarbocyanine dye, which is usually made as a perchlorate salt. Mostly used for scientific staining purposes, such as single molecule imaging, fate mapping and neuronal tracing (as DiI is retained in the lipid bilayers).

Application

1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate is used for the following applications:
  • Preparation of Dil solution
  • fluorescent membrane dye
  • Di-I Staining

Biochem/physiol Actions

Lipophilic carbocyanine dye used primarily for optical recordings of membrane voltage and for studies of membrane fluidity. Retrograde stain for neurons; provides intense, long-lasting staining of live neurons in vivo and in vitro.

Other Notes

Lipophilic fluorescent probe for studying membranes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calorimetric investigation of the phase partitioning of the fluorescent carbocyanine probes in phosphatidylcholine bilayers.
M F Ethier et al.
Biochemistry, 22(5), 1178-1182 (1983-03-01)
Yong N Li et al.
The Journal of comparative neurology, 520(16), 3786-3802 (2012-08-22)
Bipolar cells convey luminance, spatial, and color information from photoreceptors to amacrine and ganglion cells. We studied the photoreceptor connectivity of 321 bipolar cells in the adult zebrafish retina. 1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate (DiI) was inserted into whole-mounted transgenic zebrafish retinas to
Ralph Michael et al.
Vision research, 48(4), 626-634 (2008-01-29)
We evaluated the gross morphology, location, and fiber cell architecture of equatorial cortical opacities in the aging human lens. Using dark-field stereomicroscopy, we photographed donor lenses in toto and as thick slices. In addition, we investigated the details of the
R D Klausner et al.
Biochemistry, 19(26), 6199-6203 (1980-12-23)
We have examined the phase partition preferences of the even chain length (n = 10-22) diacyl-3'3'-indocarbo-cyanine iodides (Cn diI) incorporated in disaturated lecithin (PC) vesicles. Two parameters were used to determine this phase preference: (i) the direction of shift of
Muhammad Umar Jawad et al.
PloS one, 5(12), e14250-e14250 (2010-12-21)
In carcinomas stromal cells participate in cancer progression by producing proteases such as MMPs. The expression MMP1 is a prognostic factor in human chondrosarcoma, however the role in tumor progression is unknown. Laser capture microdissection and In Situ hybridization were

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service