Skip to Content
Merck
All Photos(1)

Key Documents

CLS4680

Corning® 96 Well Half-Area Microplate

High Content Imaging Film bottom, sterile, lid

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24
material:
clear bottom
cyclic olefin copolymer (COC) (COC)
flat wells
well D:
10.7 mm
well volume:
200 μL
wells:
96 wells , half area
Pricing and availability is not currently available.

material

clear bottom
cyclic olefin copolymer (COC) (COC)
flat wells

sterility

sterile

feature

lid
skirt
High Content Imaging Film bottom

packaging

case of 16 ea
pack of 4 ea

manufacturer/tradename

Corning 4680

well D

10.7 mm

well volume

200 μL

wells

96 wells , half area

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Application

Corning® 96 Well Half-Area Microplates are suitable for high-resolution cellular imaging applications.

Features and Benefits

  • Microplates made of cyclic olefin copolymer (COC) material are chemical resistant (including DMSO) and have high mechanical stability
  • Microplates with ultra-clear film of 127 μm thickness and inter- and intra-well film bottom flatness within 50 μm and 10 μm, respectively, optimized for high content applications
  • Low auto-fluorescence and birefringence

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

A frequently asked questions guide for users of the Corning® high content screening microplates.

A frequently asked questions guide for users of the Corning® high content screening microplates.

A frequently asked questions guide for users of the Corning® high content screening microplates.

A frequently asked questions guide for users of the Corning® high content screening microplates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service