Skip to Content
Merck
All Photos(1)

Key Documents

P8991

Sigma-Aldrich

Nunc® MicroWell 96 well optical bottom plates

96 well optical bottom plate, cell cultured treated, 400uL/well, black, 30/cs

Synonym(s):

96 multiwell plate, 96 well ELISA plate, 96 well immunoassay plate, 96 well microplate, 96 well microtiter plate, 96 well optical bottom plate, 96 well plate, ELISA plate, immunoassay plate, optical bottom plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
black polystyrene
flat bottom clear wells
size:
96 wells
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid
polystyrene/polymer base
Pricing and availability is not currently available.

material

black polystyrene
flat bottom clear wells

sterility

sterile

feature

lid
polystyrene/polymer base

packaging

case of 30 ea (internal packs of 10)

manufacturer/tradename

Nunc 165305

plate L × W

128 mm × 86 mm

size

96 wells

well volume

200 μL

well working volume

400 μL

working volume

200 μL

suitability

suitable for (fluorescence/luminescene based assays)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Polystyrene/Polymer Base

Optimum clarity for viewing well contents. Polymer bottom plates combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications. Polystyrene upper structures available in black or white. Flat bottom well geometry for plate reader access. Black plates provide minimum back-scatterd light. White plates maximize reflection and provide minimum autoflorescence and autoluminescence. Note: All plates supplied with lid and sterile.

White or black upper structure with polystyrene/polymer bottom plates combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications

  • Nunclon® Δ (cell culture surface), Poly-D-Lysine or Collagen I surfaces for cell culture
  • Non-treated plates are optimized for scintillation counting
  • Working volume range: 50 - 200 μl/well

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camille Dejos et al.
Scientific reports, 8(1), 5944-5944 (2018-04-15)
For many neurodegenerative disorders, expression of a pathological protein by one cell type impedes function of other cell types, which in turn contributes to the death of the first cell type. In transgenic mice modelling Stargardt-like (STGD3) maculopathy, human mutant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service