Key Documents
CLS4518
Corning® High Content Imaging Microplate
black cyclic olefin copolymer (COC), flat bottom, sterile, film bottom, lid, pack of 20
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material:
black cyclic olefin copolymer (COC)
clear bottom
flat bottom
flat bottom wells
size:
384 well
surface area:
0.03 cm2 , cell growth area
well volume:
80 μL
well working volume:
45 μL
material
black cyclic olefin copolymer (COC)
clear bottom
flat bottom
flat bottom wells
sterility
sterile
feature
lid
film bottom
low base
packaging
pack of 20 plates
pack of 20
manufacturer/tradename
Corning 4518
size
384 well
surface area
0.03 cm2 , cell growth area
well volume
80 μL
well working volume
45 μL
binding type
Tissue Culture (TC)-treated surface
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
A frequently asked questions guide for users of the Corning® high content screening microplates.
A frequently asked questions guide for users of the Corning® high content screening microplates.
A frequently asked questions guide for users of the Corning® high content screening microplates.
A frequently asked questions guide for users of the Corning® high content screening microplates.