Skip to Content
Merck
All Photos(1)

Key Documents

A9HT

88042804, mouse unknown/unspecified, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

A9HT, 88042804

biological source

mouse unknown/unspecified

packaging

tube of 5 μg 88042804-DNA-5UG
pkg of vial of cells 88042804-1VL

growth mode

Adherent

karyotype

Hyperdiploid

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

A9 tumour induced C3H mouse

Cell Line Description

This cell line was selected by passaging A9 (Sigma Catalogue number. 85011426) through syngenic C3H or semi allogenic mice over a period of three months and uptake was considerably higher than A9, thus A9 high uptake or A9HT. Karyotype is similar to A9 but with a slightly reduced total number of chromosomes, with the modal number of 53. This line lacks inosinic acid pyrophosphorylase.

Application

Tumourigenicity studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service