Skip to Content
Merck
All Photos(1)

Key Documents

10812846001

Roche

Tris hydrochloride

>99% (titration), pH 7.0-9.0, suitable for FISH

Synonym(s):

Trizma® hydrochloride, TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
Beilstein:
3675235
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

Assay

>99% (titration)

form

solid

packaging

pkg of 500 g

manufacturer/tradename

Roche

technique(s)

FISH: suitable

pH

7.0-9.0

useful pH range

7.0-9.0

pKa (25 °C)

8.1

absorption

<0.02 at 300 nm at 100 mg/mL

storage temp.

room temp

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Buffering agent in incubation mixtures. It has also been used as a component of lysis and TE (Tris-EDTA) buffer.

Features and Benefits

Crystals

Quality

Purity: >99% Tris-HCl (titrimetric)
Heavy metals: <1 ppm Pb and Fe

Analysis Note

Purity: >99% Tris-HCl (titrimetric)
Heavy metals: <1 ppm Pb and Fe

Other Notes

The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution. For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MNase Digestion for Nucleosome Mapping in Neurospora.
Cigdem S
Bio-protocol null
Izabela Jęśkowiak et al.
International journal of molecular sciences, 22(20) (2021-10-24)
(1) Background: Melanoma is an aggressive neoplasm derived from melanocyte precursors with a high metastatic potential. Responses to chemotherapy and immunotherapy for melanoma remain weak, underlining the urgent need to develop new therapeutic strategies for the treatment of melanoma. (2)
Cynthia Vanesa Rivero et al.
Frontiers in cellular and infection microbiology, 11, 657257-657257 (2021-09-04)
T. cruzi, the causal agent of Chagas disease, is a parasite able to infect different types of host cells and to persist chronically in the tissues of human and animal hosts. These qualities and the lack of an effective treatment
Alessio Fragasso et al.
ACS nano (2021-06-26)
Molecular traffic across lipid membranes is a vital process in cell biology that involves specialized biological pores with a great variety of pore diameters, from fractions of a nanometer to >30 nm. Creating artificial membrane pores covering similar size and
Magalí Martí et al.
STAR protocols, 2(3), 100652-100652 (2021-07-27)
16S rRNA gene sequencing enables microbial community profiling, but recovering fecal DNA from extremely premature infants is challenging. Here, we describe an optimized protocol for fecal DNA isolation, library preparation for 16S rRNA gene sequencing, taxonomy assignation, and statistical analyses.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service