Skip to Content
Merck
All Photos(1)

Documents

12-303

Sigma-Aldrich

Jurkat Cell Lysate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105330
eCl@ss:
32011807
NACRES:
NA.41

biological source

human

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

shipped in

dry ice

Cell Line Description

Jurkat

Quality

Routinely evaluated as a positive control for some of Upstates antibodies.

Physical form

Modified RIPA buffer (50mM Tris-HCl, pH 7.4, 1% NP40, 0.25% sodium deoxycholate, 150mM NaCl, 1mM EDTA, 1mM PMSF, 1mg/ml aprotinin, 1mg/ml leupeptin, 1mg/ml pepstatin, 1mM Na3VO4, 1mM NaF) and diluted with non-reducing sample buffer (31mM Tris-HCl, pH 6.8, 5% glycerol, 1% SDS, 0.002% bromphenol blue).

Storage and Stability

6 months at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peixuan Zhu et al.
Leukemia & lymphoma, 54(3), 579-586 (2012-07-31)
Overexpression of zeta-chain-associated protein 70 (ZAP-70) was recently recognized as an independent prognostic marker for the aggressive form of chronic lymphocytic leukemia (CLL). The objective of this study was to demonstrate the feasibility and implementation of quantitative detection of ZAP-70
Xavier Le Guezennec et al.
Molecular and cellular biology, 26(3), 843-851 (2006-01-24)
The human genome contains a number of methyl CpG binding proteins that translate DNA methylation into a physiological response. To gain insight into the function of MBD2 and MBD3, we first applied protein tagging and mass spectrometry. We show that
Gianluca Rosso et al.
International journal of bipolar disorders, 11(1), 33-33 (2023-10-09)
The differential diagnosis of patients presenting for the first time with a depressive episode into unipolar disorder versus bipolar disorder is crucial to establish the correct pharmacological therapy (antidepressants vs mood stabilizers), but no biological markers are currently available. Several
Apostolos Polykratis et al.
The Journal of biological chemistry, 280(23), 22454-22461 (2005-03-31)
Heparin affin regulatory peptide (HARP) is an 18-kDa secreted growth factor that has a high affinity for heparin and a potent role on tumor growth and angiogenesis. We have previously reported that HARP is mitogenic for different types of endothelial
Cori N Booker et al.
NPJ microgravity, 7(1), 49-49 (2021-11-28)
Disuse osteoporosis (DO) results from mechanical unloading of weight-bearing bones and causes structural changes that compromise skeletal integrity, leading to increased fracture risk. Although bone loss in DO results from imbalances in osteoblast vs. osteoclast activity, its effects on skeletal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service