Skip to Content
Merck
All Photos(2)

Documents

C7081

Sigma-Aldrich

WY-14643

Synonym(s):

4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthioacetic acid, Pirinixic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14ClN3O2S
CAS Number:
Molecular Weight:
323.80
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

SMILES string

CC1=C(C)C=CC=C1NC2=NC(SCC(O)=O)=NC(Cl)=C2

InChI

1S/C14H14ClN3O2S/c1-8-4-3-5-10(9(8)2)16-12-6-11(15)17-14(18-12)21-7-13(19)20/h3-6H,7H2,1-2H3,(H,19,20)(H,16,17,18)

InChI key

SZRPDCCEHVWOJX-UHFFFAOYSA-N

Gene Information

General description

WY-14643 prevents lipopolysaccharide (LPS)-induced inflammation in synovial fibroblasts. It protects cortical neurons from pro-inflammatory mediator-induced cell injuries. WY-14643 has inhibitory effects on pro-inflammatory responses in microglia. It improves oxido-nitrosative stress in disease models and reduces the generation of reactive oxygen species (ROS), nitric oxide (NO) and lipid peroxidation end-products in the brain.

Application

WY-14643 has been used:
  • as a positive control for transfection and luciferase assay
  • to stimulate bone marrow–derived macrophages (BMDM) for autophagic flux analysis
  • to analyze the regulation of fatty acid metabolism-immune nexus (FAMIN) expression in cell culture

Biochem/physiol Actions

Selective PPARα agonist.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) and Nuclear Receptors (PPARs) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joe Jongpyo Lim et al.
Drug metabolism and disposition: the biological fate of chemicals, 50(10), 1396-1413 (2021-12-04)
Perfluorinated carboxylic acids (PFCAs) are widespread environmental pollutants for which human exposure has been documented. PFCAs at high doses are known to regulate xenobiotic transporters partly through peroxisome proliferator-activated receptor alpha (PPARα) and constitutive androstane receptor (CAR) in rodent models.
Vladimir S Shavva et al.
Molecular and cellular biochemistry, 448(1-2), 211-223 (2018-02-15)
Apolipoprotein A-I (ApoA-I) is the main structural and functional protein component of high-density lipoprotein. ApoA-I has been shown to regulate lipid metabolism and inflammation in macrophages. Recently, we found the moderate expression of endogenous apoA-I in human monocytes and macrophages
Stefan Clockaerts et al.
Annals of the rheumatic diseases, 71(6), 1012-1018 (2012-02-07)
Infrapatellar fat pad (IPFP) might be involved in osteoarthritis (OA) by production of cytokines. It was hypothesised that production of cytokines is sensitive to environmental conditions. To evaluate cytokine production by IPFP in response to interleukin (IL)1β and investigate the
Ronald Lucarelli et al.
Frontiers in cellular and infection microbiology, 12, 881462-881462 (2022-07-22)
Secondary bacterial infection (superinfection) post influenza is a serious clinical complication often leading to pneumonia and death. Eicosanoids are bioactive lipid mediators that play critical roles in the induction and resolution of inflammation. CYP450 lipid metabolites are anti-inflammatory lipid mediators
Two Isomeric C16 Oxo-Fatty Acids from the Diatom Chaetoceros karianus Show Dual Agonist Activity towards Human Peroxisome Proliferator-Activated Receptors (PPARs) alphagamma
Moldes-Anaya A, et al.
Marine drugs, 15(6), 148-148 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service