AXYT200CLRS
MAXYMum Recovery® pipette tips
racked, size 200 μL, clear, pkg of 5x960tips/cs, sterile
Synonym(s):
Pipette tips, pipette tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
volume:
200 μL
sterility:
sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter: no
RNase free
barrier: no
endotoxin free
filter: no
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc T-200-C-L-R-S
Corning
Corning
Recommended Products
material
clear
sterility
sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter: no
manufacturer/tradename
Axygen Inc T-200-C-L-R-S
Corning
packaging
pkg of 5x960tips/cs
size
200 μL
volume
200 μL
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
Axygen® MAXYMum Recovery® pipette tips provide the following benefits:
- Tips fit a broad range of single and multi-channel pipettors
- Compatible with most of the well-known brands of pipettors
- Tips are made of polypropylene and are non-pyrogenic
- Tips are RNAse and DNAse free
- Flexible walls and a series of internal sealing rings provides good fitting which requires less force to load and eject
- Beveled
- Low-risk of sample loss due to high fluid retention
- Low-risk of cross-contamination
- Higher accuracy and precision
Other Notes
Browse all pipette tips
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Maxymum Recovery is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service