Skip to Content
Merck
All Photos(1)

Key Documents

MABS496

Sigma-Aldrich

Anti-Mineralocorticoid Receptor Antibody, clone 6G1

clone 6G1, from mouse

Synonym(s):

MR, Nuclear receptor subfamily 3 group C member 2, Mineralocorticoid Receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6G1, monoclonal

species reactivity

rat, mouse

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Nr3C2(110784)

General description

Mineralocorticoid receptor (UniProt P22199; also known as MR, Nuclear receptor subfamily 3 group C member 2) is encoded by the Nr3c2 (also known as MCR, Mlr) gene (Gene ID 25672) in rats. Mineralocorticoid receptor (MR) is expressed in multiple tissues, including renal epithelia, smooth muscle, endothelium, cardiomyocytes, and hippocampal neurons, where it mediates diverse functions. MR is activated by aldosterone and also by cortisol in cells that do not express 11β-hydroxysteroid dehydrogenase type 2 (11βHSD2). MR is normally activated by aldosterone, which is produced by adrenal glomerulosa in response to intravascular volume depletion and hyperkalemia. Consistently, gain of function mutations in MR lead to severe hypertension, often with hypokalemia, while loss of function mutations result in neonatal hypotension. Ser843 phosphorylation in the MR ligand-binding domain is reported to prevent MR ligand binding and activation. MR pS843 is found exclusively in intercalated cells of the distal nephrons in the kidney. Intravascular volume depletion, angiotensin II and WNK4 signaling reduce MR pS843 levels, whereas hyperkalemia increases MR pS843.

Specificity

Epitope is present in all 3 alternative spliced isoforms.

Immunogen

Conjugated linear peptide corresponding to rat Mineralocorticoid Receptor.

Application

Detect Mineralocorticoid Receptor using this Anti-Mineralocorticoid Receptor Antibody, clone 6G1 validated for use in Western Blotting, Immunohistochemistry and Immunocytochemistry.
Immunohistochemistry Analysis: A representative lot detected Mineralocorticoid Receptor in various rat tissues including kidney, hippocampus, choroid plexus, cerebellum, colon, heart, and coronary artery (Gomez-Sanchez, C.E., et al. (2006). Endocrinology. 147(3):1343-1348).
Immunocytochemistry Analysis: A representative lot detected Mineralocorticoid Receptor in mouse kidney cells (Shibata, S., et al. (2013). Cell Metabolism. 18:660-671).
Western Blotting Analysis: A representative lot detected Mineralocorticoid Receptor in rat hippocampal tissue lysate (Gomez-Sanchez, C.E., et al. (2006). Endocrinology. 147(3):1343-1348).
Western Blotting Analysis: A representative lot detected endogenous Mineralocorticoid Receptor (MR) in mouse kidney and exogenously expressed human MR in transfected COS-7 cells (Shibata, S., et al. (2013) Cell Metab. 18(5):660-671).

Quality

Evaluated by Western Blotting in M1MR cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Mineralocorticoid Receptor in 10 µg of M1MR cell lysate.

Target description

~107 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoual Dahmana et al.
International journal of pharmaceutics, 604, 120773-120773 (2021-06-07)
Impaired wound healing in patients receiving glucocorticoid therapy is a serious clinical concern: mineralocorticoid receptor (MR) antagonists can counter glucocorticoid-induced off-target activation of MR receptors. The aim of this study was to investigate the cutaneous delivery of the potent MR
Viren H Makhijani et al.
Neuropharmacology, 181, 108337-108337 (2020-10-03)
The mineralocorticoid receptor (MR) is an emerging target in the field of alcohol research. The MR is a steroid receptor in the same family as the glucocorticoid receptor, with which it shares the ligand corticosterone in addition to the MR
Mahshid Gazorpak et al.
Nature communications, 14(1), 8177-8177 (2023-12-10)
Counteracting the overactivation of glucocorticoid receptors (GR) is an important therapeutic goal in stress-related psychiatry and beyond. The only clinically approved GR antagonist lacks selectivity and induces unwanted side effects. To complement existing tools of small-molecule-based inhibitors, we present a
Serena M Dudek et al.
Hippocampus, 33(6), 700-711 (2023-05-09)
Since 1959, the Russian Farm-Fox study has bred foxes to be either tame or, more recently, aggressive, and scientists have used them to gain insight into the brain structures associated with these behavioral features. In mice, hippocampal area CA2 has
Jakob Hartmann et al.
Cell reports, 35(9), 109185-109185 (2021-06-03)
Responding to different dynamic levels of stress is critical for mammalian survival. Disruption of mineralocorticoid receptor (MR) and glucocorticoid receptor (GR) signaling is proposed to underlie hypothalamic-pituitary-adrenal (HPA) axis dysregulation observed in stress-related psychiatric disorders. In this study, we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service