Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MAXYMum Recovery
®
pipette tips
Products
Product Category
pipette tips (12)
Manufacturer
Color
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
AXYT200CLRS
MAXYMum Recovery
®
pipette tips,
racked, size 200 μL, clear, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYT300LRS
MAXYMum Recovery
®
pipette tips,
for P2/P10, racked, size 0.5-10 μL, pkg of 5x960tips/cs, clear, sterile
Expand
View Pricing
AXYT300LR
MAXYMum Recovery
®
pipette tips,
for P2/P10, racked, size 0.5-10 μL, pkg of 5x960tips/cs, clear
Expand
View Pricing
AXYT1000CL
MAXYMum Recovery
®
pipette tips,
capacity 1000 μL, clear, pkg of 5x1000tips/cs
Expand
View Pricing
AXYT200CLR
MAXYMum Recovery
®
pipette tips,
racked, size 200 μL, clear, pkg of 5x960tips/cs
Expand
View Pricing
AXYT1000CLRS
MAXYMum Recovery
®
pipette tips,
racked, capacity 1000 μL, clear, pkg of 5x1000tips/cs, sterile
Expand
View Pricing
AXYT300LSTK
MAXYMum Recovery
®
pipette tips,
for P2/P10 in stacked racks, size 0.5-10 μL, pkg of 10x480tips/cs, clear
Expand
View Pricing
AXYT350CLR
MAXYMum Recovery
®
pipette tips,
racked, size 300 μL, clear, pkg of 5x960tips/cs
Expand
View Pricing
AXYT350CL
MAXYMum Recovery
®
pipette tips,
size 300 μL, clear, pkg of 10x1000tips/cs
Expand
View Pricing
AXYT1000CLR
MAXYMum Recovery
®
pipette tips,
racked, capacity 1000 μL, clear, pkg of 5x1000tips/cs
Expand
View Pricing
AXYT200CLSTK
MAXYMum Recovery
®
pipette tips,
stacked racks, size 200 μL, clear, pkg of 10x480tips/cs
Expand
View Pricing
AXYT350CLRS
MAXYMum Recovery
®
pipette tips,
racked, size 300 μL, clear, pkg of 5x960tips/cs, sterile
Expand
View Pricing