Skip to Content
Merck
All Photos(1)

Key Documents

A9920

Sigma-Aldrich

β-Alanine

BioReagent, suitable for cell culture, suitable for insect cell culture

Synonym(s):

β-Ala, 3-Aminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2COOH
CAS Number:
Molecular Weight:
89.09
Beilstein:
906793
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
39030936
PubChem Substance ID:
NACRES:
NA.75

product line

BioReagent

Assay

≥98% (TLC)

form

powder or crystals

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

color

white

mp

202 °C (dec.) (lit.)

SMILES string

NCCC(O)=O

InChI

1S/C3H7NO2/c4-2-1-3(5)6/h1-2,4H2,(H,5,6)

InChI key

UCMIRNVEIXFBKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

An N-blocked form of alanine.

Biochem/physiol Actions

Endogenous β-amino acid that is a nonselective agonist at glycine receptors and a ligand for the G protein-coupled orphan receptor, TGR7 (MrgD). β-Alanine is a component of pantothenic acid.
β-Alanine, a β−amino acid, is a component of pantothenic acid and the rate-limiting amino acid in the biosynthesis of the histidinyl antioxidant dipeptides carnosine and anserine. Endogenous β-amino acid that is a nonselective agonist at glycine receptors and a ligand for the G protein-coupled orphan receptor, TGR7 (MrgD). β-Alanine flux plays a cytoprotective role by supporting the osmotic stability of marine organisms, preimplantation mouse embryos and mammalian cells exposed to hypoxic stress.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert G Hart et al.
The Canadian journal of cardiology, 29(7 Suppl), S71-S78 (2013-06-29)
Chronic kidney disease (CKD) is prevalent in elderly patients with atrial fibrillation and is an independent risk factor for stroke. Warfarin anticoagulation is efficacious for stroke prevention in atrial fibrillation patients with moderate CKD (stage III, estimated glomerular filtration rate
K M Gibson et al.
Journal of neurochemistry, 81(1), 71-79 (2002-06-18)
Metabolite profiling in succinate semialdehyde dehydrogenase (SSADH; Aldh5a1-/-) deficient mice previously revealed elevated gamma-hydroxybutyrate (GHB) and total GABA in urine and total brain and liver extracts. In this study, we extend our metabolic characterization of these mutant mice by documenting
Partha Sardar et al.
The American journal of cardiology, 113(7), 1173-1177 (2014-02-12)
Dabigatran is a novel oral anticoagulant and may be useful during atrial fibrillation (AF) ablation for prevention of thromboembolic events. However, the benefits and adverse effects of periprocedural dabigatran therapy have not been thoroughly evaluated. A meta-analysis was performed to
Julie Y Culbertson et al.
Nutrients, 2(1), 75-98 (2010-01-01)
Muscle carnosine has been reported to serve as a physiological buffer, possess antioxidant properties, influence enzyme regulation, and affect sarcoplasmic reticulum calcium regulation. Beta-alanine (β-ALA) is a non-essential amino acid. β-ALA supplementation (e.g., 2-6 grams/day) has been shown to increase
Robert B Campenot et al.
Nature protocols, 4(12), 1869-1887 (2009-12-17)
The compartmented culture, in which primary neurons plated in a proximal compartment send their axons under silicone grease barriers and into left and right distal compartments, has enhanced the experimental capabilities of neuronal cultures. Treatments can be applied separately to

Articles

Learn more about β-Alanine, an important amino acid supplement and explore how it is used in cell culture. We offer a comprehensive portfolio of amino acid cell culture supplements and reagents.

Learn more about β-Alanine, an important amino acid supplement and explore how it is used in cell culture. We offer a comprehensive portfolio of amino acid cell culture supplements and reagents.

Learn more about β-Alanine, an important amino acid supplement and explore how it is used in cell culture. We offer a comprehensive portfolio of amino acid cell culture supplements and reagents.

Learn more about β-Alanine, an important amino acid supplement and explore how it is used in cell culture. We offer a comprehensive portfolio of amino acid cell culture supplements and reagents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service