Skip to Content
Merck
All Photos(3)

Key Documents

A5502

Sigma-Aldrich

Adonitol

≥99%

Synonym(s):

Adonite, Ribitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O5
CAS Number:
Molecular Weight:
152.15
Beilstein:
1720524
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥99%

form

powder

color

white

mp

104  °C ((219 °F ))

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

SMILES string

OC[C@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4+,5-

InChI key

HEBKCHPVOIAQTA-ZXFHETKHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Adonitol (Ribitol), a pentose alcohol, is metabolized to teicholic acids used in the cell walls of gram positive bacteria. Adonitol is often compared to other cell permeating molecules such as formamide, propanediol, and DMSO as a cryopreservation agent.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anders Østergaard Madsen et al.
The journal of physical chemistry. A, 115(26), 7794-7804 (2011-06-15)
X-ray diffraction data of high quality measured to high resolution on crystals of the two pentitol epimers ribitol (centric) and xylitol (acentric) at 101, 141, and 181 K and data on the two compounds previously recorded at 122 K have
Annelies Goeminne et al.
Bioorganic & medicinal chemistry, 16(14), 6752-6763 (2008-06-24)
A key enzyme within the purine salvage pathway of parasites, nucleoside hydrolase, is proposed as a good target for new antiparasitic drugs. We have developed N-arylmethyl-iminoribitol derivatives as a novel class of inhibitors against a purine specific nucleoside hydrolase from
S H Moolenaar et al.
NMR in biomedicine, 14(3), 167-176 (2001-05-18)
In vivo NMR spectroscopy was performed on the brain of a patient with a leukoencephalopathy, revealing unknown resonances between 3.5 and 4.0 ppm. In addition, urine and CSF of the patient were measured using high-resolution NMR spectroscopy. Also in these
Claudia Bello et al.
Bioorganic & medicinal chemistry, 19(24), 7720-7727 (2011-11-15)
New derivatives of 1,4-dideoxy-1,4-imino-D-ribitol have been prepared and evaluated for their cytotoxicity on solid and haematological malignancies. 1,4-Dideoxy-5-O-[(9Z)-octadec-9-en-1-yl]-1,4-imino-D-ribitol (13, IC(50) ∼2 μM) and its C(18)-analogues (IC(50) <10 μM) are cytotoxic toward SKBR3 (breast cancer) cells. 13 also inhibits (IC(50) ∼8
Alec R Nickolls et al.
Disease models & mechanisms, 13(6) (2020-05-20)
The basal lamina is a specialized sheet of dense extracellular matrix (ECM) linked to the plasma membrane of specific cell types in their tissue context, which serves as a structural scaffold for organ genesis and maintenance. Disruption of the basal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service