Skip to Content
Merck
All Photos(1)

Key Documents

146064

Sigma-Aldrich

β-Alanine

99%, suitable for ligand binding assays

Synonym(s):

β-Ala, 3-Aminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2COOH
CAS Number:
Molecular Weight:
89.09
Beilstein:
906793
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
39030936
PubChem Substance ID:
NACRES:
NA.26

Product Name

β-Alanine, 99%

Assay

99%

form

powder, crystals or granules

technique(s)

ligand binding assay: suitable

color

white

mp

202 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

NCCC(O)=O

InChI

1S/C3H7NO2/c4-2-1-3(5)6/h1-2,4H2,(H,5,6)

InChI key

UCMIRNVEIXFBKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

β-Alanine has been used as a ligand for the orphan MAS-related receptor. It has also been used in culture media used for certain strains of yeast to test for β-alanine auxotrophy.

Biochem/physiol Actions

Endogenous β-amino acid that is a nonselective agonist at glycine receptors and a ligand for the G protein-coupled orphan receptor, TGR7 (MrgD). β-Alanine is a component of pantothenic acid.
β-Alanine, a β−amino acid, is a component of pantothenic acid and the rate-limiting amino acid in the biosynthesis of the histidinyl antioxidant dipeptides carnosine and anserine. Endogenous β-amino acid that is a nonselective agonist at glycine receptors and a ligand for the G protein-coupled orphan receptor, TGR7 (MrgD). β-Alanine flux plays a cytoprotective role by supporting the osmotic stability of marine organisms, preimplantation mouse embryos and mammalian cells exposed to hypoxic stress.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saccharomyces cerevisiae is capable of de Novo pantothenic acid biosynthesis involving a novel pathway of beta-alanine production from spermine.
White WH
The Journal of Biological Chemistry, 276(14), 10794-10800 (2001)
The orphan G protein-coupled receptor GPR139 is activated by the peptides: Adrenocorticotropic hormone (ACTH), a-, and ?-melanocyte stimulating hormone (a-MSH, and ?-MSH), and the conserved core motif HFRW.
N?hr AC, et al.
Neurochemistry International, 102, 105-113 (2017)
Ramin Artang et al.
The American journal of cardiology, 112(12), 1973-1979 (2013-10-01)
Dabigatran has been associated with greater risk of myocardial infarction (MI) than warfarin. It is unknown whether the increased risk is unique to dabigatran, an adverse effect shared by other oral direct thrombin inhibitors (DTIs), or the result of a
Ammar Majeed et al.
Circulation, 128(21), 2325-2332 (2013-10-02)
The aim of this study was to compare the management and prognosis of major bleeding in patients treated with dabigatran or warfarin. Two independent investigators reviewed bleeding reports from 1034 individuals with 1121 major bleeds enrolled in 5 phase III
Robert G Hart et al.
The Canadian journal of cardiology, 29(7 Suppl), S71-S78 (2013-06-29)
Chronic kidney disease (CKD) is prevalent in elderly patients with atrial fibrillation and is an independent risk factor for stroke. Warfarin anticoagulation is efficacious for stroke prevention in atrial fibrillation patients with moderate CKD (stage III, estimated glomerular filtration rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service