Skip to Content
Merck
All Photos(7)

Key Documents

A8763

Sigma-Aldrich

Albumin from human serum

lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)

Synonym(s):

HSA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42010201
NACRES:
NA.25

biological source

human

Assay

≥99% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

monomer calculated mol wt 66478 Da

concentration

≤1% (Globulin)

technique(s)

ELISA: suitable
tissue culture: suitable
western blot: suitable

impurities

HIV I and HIVII, HCV and HBsAg, tested negative

solubility

H2O: soluble 20 mg/mL

UniProt accession no.

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

Gene Information

human ... ALB(213)

Looking for similar products? Visit Product Comparison Guide

General description

Albumin is the most copious protein in blood plasma. Liver produces human albumin.

Application

Albumin was used to test its effect on the in vitro bactericidal activity of cefditoren against penicillin-resistant Streptococcus pneumonia. It has been clinically used in serious and often life-threatening conditions, such as shock and blood loss due to trauma, burns, and surgery.
Albumin was used to study albumin-induced apoptosis in proximal tubule cells. It was also used in cell adhesion molecules and surface treatment studies.

Biochem/physiol Actions

Albumins are soluble monomeric proteins found in the body fluids and tissues of animals and in some plant seeds. Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. Serum albumins are also vital in regulating the colloidal osmotic pressures of blood.
Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Albumin can also bind to exogenous substances, particularly drugs (e.g., ibuprofen, warfarin), and strongly influences their pharmacokinetics. Oxidative stress leading to changes in the redox state of albumin has varied effects on its physiological function.
Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Albumin is also seen to bind to exogenous substances, particularly drugs (e.g., ibuprofen, warfarin), and strongly influence their pharmacokinetics. Oxidative stress leading to changes in the redox state of albumin has widely varied effects on its physiological function.

Other Notes

View more information on human serum albumin.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamás Pivarcsik et al.
Pharmaceuticals (Basel, Switzerland), 14(6) (2021-06-03)
In this work, the various biological activities of eight organoruthenium(II) complexes were evaluated to reveal correlations with their stability and reactivity in aqueous media. Complexes with general formula [Ru(η6-p-cymene)(X,Y)(Z)] were prepared, where (X,Y) represents either an O,O-ligand (β-diketone), N,O-ligand (8-hydroxyquinoline)
H K Parmentier et al.
Poultry science, 88(3), 536-542 (2009-02-13)
To mimic airborne immune challenges, layer hens were intratracheally and concurrently challenged with various doses of the protein antigen human serum albumin (HuSA) and the pathogen-associated molecular pattern lipopolysaccharide (LPS) at 7 and 13 wk of age. All groups received
Mateusz P Czub et al.
Molecular pharmacology, 98(6), 648-657 (2020-09-27)
Protein tyrosine phosphatase (PTP) 4A3 is frequently overexpressed in human solid tumors and hematologic malignancies and is associated with tumor cell invasion, metastasis, and a poor patient prognosis. Several potent, selective, and allosteric small molecule inhibitors of PTP4A3 were recently
Nóra V May et al.
Molecules (Basel, Switzerland), 26(9) (2021-06-03)
Copper(II) complexes of thiosemicarbazones (TSCs) often exhibit anticancer properties, and their pharmacokinetic behavior can be affected by their interaction with blood transport proteins. Interaction of copper(II) complexes of an {N,N,S} donor α-N-pyridyl TSC (Triapine) and an {O,N,S} donor 2-hydroxybenzaldehyde TSC
Nitin Agrawal et al.
Lab on a chip, 8(12), 2054-2061 (2008-11-22)
Neutrophil directional migration in response to chemical gradients, also known as chemotaxis, is one of the key phenomena in the immune responses against bacterial infection. To better study neutrophils chemotaxis, several in vitro assays have been developed that replicate chemotactic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service