Skip to Content
Merck
All Photos(4)

Key Documents

A9731

Sigma-Aldrich

Albumin human

Cellastim S, recombinant, expressed in rice, lyophilized powder, suitable for cell culture, low endotoxin, ≥96% (SDS-PAGE)

Synonym(s):

Cellastim S, rHSA, rHSA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2936H4624N786O889S41
CAS Number:
Molecular Weight:
66471.53
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42010201
NACRES:
NA.25

biological source

human

Quality Level

recombinant

expressed in rice

Assay

≥96% (SDS-PAGE)

form

lyophilized powder

mol wt

monomer ~67 kDa

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/mg protein

UniProt accession no.

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

Gene Information

human ... ALB(213)

Looking for similar products? Visit Product Comparison Guide

Application

This recombinant variant is recommended for use in absorption, distribution, metabolism, and excretion (ADME) pharmacological research; cell culture; drug delivery research; and cryopreservation of cells.

Biochem/physiol Actions

Albumins are soluble monomeric proteins found in the body fluids and tissues of animals and in some plant seeds. Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. Serum albumins are also vital in regulating the colloidal osmotic pressures of blood.

Features and Benefits

Cellastim is a well defined, animal-free recombinant human albumin (rHA). Designed for cell culture applications, Cellastim is a powerful media supplement that outperforms plasma-derived human albumin, bovine serum albumin, and other sources of recombinant human albumin. Cellastim delivers consistent performance, not the variability associated with hydrolysates and extracts. Cellastim is ideally suited for use in cell culture media for Chinese Hamster Ovary (CHO), hybridoma, VERO, stem cells and primary cells.

Advantages
  • Eliminates animal-derived additives
  • Safe and regulatory friendly
  • Proven effective in enhancing stem cell growth in embryonic, mesenchymal, neural, cardiac, liver, kidney, keratinocyte and fibroblast stem cell lines
  • Maintains stem cells in an undifferentiated state
  • Eliminates coating of culture plates for adherent cell lines
  • Provides well-defined and consistent stem cell medium

Analysis Note

Assayed by SDS-PAGE

Other Notes

1). Patent Rights
EP2230311
U.S. Patent No. 10,618,951
U.S. Patent No. 10,981,974
U.S. Patent No. 11,492,3892.
2). Registered Trademark Rights
U.S. Reg. 5,696,531
International Reg. 1,456,207
To view more information on Cellastim download the Cellastim Brochure, Cellastim CHO Poster, and Cellastim hybridoma Poster.
View more information on human serum albumin.

Legal Information

Cellastim is a trademark of InVitria

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junfeng Zhu et al.
Journal of pharmaceutical and biomedical analysis, 134, 100-107 (2016-11-28)
Danhong injection (DHI) is a widely used Chinese medicine injection (CMI) for the clinical treatment of cardiovascular and cerebrovascular diseases. In this study, a simple and efficient in vitro method based on ultrafiltration LC-MS and molecular modeling has been developed
Wireeya Chawjiraphan et al.
MethodsX, 7, 101114-101114 (2020-11-17)
A simple and sensitive graphene oxide-mediated fluorescence quenching aptasensor is developed to quantify albuminuria in urine samples. The developed aptasensor used the specific target binding property of aptamer and fluorescence quenching property of graphene oxide to determine the concentration of
Yasuhiro Kawai et al.
PloS one, 13(5), e0196655-e0196655 (2018-05-26)
Serum albumin is the major intravascular antioxidant. Though oxidative stress plays an important role in the pathophysiology of Immunoglobulin A nephropathy (IgAN), the association between serum albumin and the progression of IgAN is not entirely understood. This retrospective cohort study
Yongjia Fan et al.
ACS biomaterials science & engineering, 3(8), 1510-1518 (2016-08-14)
The development of platforms for the expansion and directed differentiation of human pluripotent stem cells (hPSCs) in large quantities under xeno-free conditions is a key step toward the realization of envisioned stem cell-based therapies. Microcarrier bioreactors afford great surface-to-volume ratio
D Moreno et al.
Theriogenology, 84(7), 1053-1060 (2015-08-19)
The aim of this study was to develop an in vitro embryo culture medium without either fetal calf serum or BSA, using various growth factors and cytokines (GFs-CYKs; IGF-I, IGF-II, bFGF, LIF, GM-CSF, TGF-β1, and PDGF-BB), and other molecules with

Articles

Discover answers to your frequently asked questions about animal component-free, xenobiotic-free, and chemically defined media and reagents for cell culture.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service