Skip to Content
Merck
All Photos(1)

Key Documents

A5843

Sigma-Aldrich

Albumin from human serum

lyophilized powder, low endotoxin

Synonym(s):

HSA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42010201
NACRES:
NA.25

biological source

human

Assay

≥96% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

monomer calculated mol wt 66478 Da

technique(s)

activity assay: suitable

impurities

HIV I and HIVII, HCV and HBsAg, tested negative
≤1.0 EU/mg endotoxin

solubility

H2O: soluble 50 mg/mL

UniProt accession no.

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

Gene Information

human ... ALB(213)

Looking for similar products? Visit Product Comparison Guide

General description

Albumin is the most copious protein in blood plasma. Liver produces human albumin.

Application

Albumin was used to test its effect on the in vitro bactericidal activity of cefditoren against penicillin-resistant Streptococcus pneumonia. It has been clinically used in serious and often life-threatening conditions, such as shock and blood loss due to trauma, burns, and surgery.
Albumin was used to study albumin-induced apoptosis in proximal tubule cells. It was also used in cell adhesion molecules and surface treatment studies.

Biochem/physiol Actions

Albumins are soluble monomeric proteins found in the body fluids and tissues of animals and in some plant seeds. Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. Serum albumins are also vital in regulating the colloidal osmotic pressures of blood.
Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Albumin is also seen to bind to exogenous substances, particularly drugs (e.g., ibuprofen, warfarin), and strongly influence their pharmacokinetics. Oxidative stress leading to changes in the redox state of albumin has widely varied effects on its physiological function.
Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Albumin is also seen to bind to exogenous substances, particularly drugs (e.g., ibuprofen, warfarin), and strongly influences their pharmacokinetics. Oxidative stress leading to changes in the redox state of albumin has varied effects on its physiological function.

Other Notes

View more information on human serum albumin.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sam Coffey et al.
PloS one, 10(10), e0140417-e0140417 (2015-10-16)
Diabetes mellitus (DM) has become an epidemic, causing a significant decline in quality of life of individuals due to its multisystem involvement. Kidney is an important target organ in DM accounting for the majority of patients requiring renal replacement therapy
Edina Korça et al.
Scientific reports, 10(1), 17105-17105 (2020-10-15)
Advanced glycation endproducts (AGEs) are formed in a series of non-enzymatic reactions between reducing sugars and the amino groups of proteins and accumulate during aging, diabetes mellitus, chronic kidney disease and other chronic diseases. Accumulation of AGE-modifications alters protein structure
Qiong Wen et al.
Nephrology (Carlton, Vic.), 15(1), 63-74 (2010-04-10)
SUMMARY AT A GLANCE Patients with MCD and FSGS had significant changes in the expression of several EMT-related urinary proteins through activation ERK1/2 and p38 signaling pathways. Further studies to explore the molecular mechanisms behind the distinct clinical features of
M van de Vyver et al.
Scandinavian journal of medicine & science in sports, 26(6), 638-647 (2015-06-11)
High-intensity exercise results in immune activation. This study determined whether (a) there is concordance between serum MPO and neutrophil and/or monocyte intracellular MPO content; (b) peripheral blood mononuclear cells respond to inflammatory interleukins (ILs) by increasing intracellular signaling. Healthy male
Pasquale Cervero et al.
Methods in molecular biology (Clifton, N.J.), 1046, 97-121 (2013-07-23)
Podosomes are multifunctional organelles of invasive cells that combine several key abilities including cell-matrix adhesion, extracellular matrix degradation, and mechanosensing. In combination with their high turnover rates that allow quick adaptation to the pericellular environment, podosomes are likely to play

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service