Skip to Content
Merck
All Photos(1)

Key Documents

A7544

Sigma-Aldrich

Anti-Albumin antibody produced in goat

whole antiserum

Synonym(s):

Serum albumin antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

15 mM sodium azide

species reactivity

human

technique(s)

indirect ELISA: 1:50,000
quantitative precipitin assay: 3.0

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALB(213)

Related Categories

General description

Albumin (ALB) is an autosomal gene with 15 exons mapped to human chromosome 4q13.3. The gene codes for a circulating human albumin protein containing 585 amino acid residues and 17 disulfide bridges. It is biosynthesized in the hepatocyte.

Immunogen

purified human albumin

Application

Anti-Albumin antibody produced in goat has been used in sandwich enzyme linked immunosorbent assays (ELISA), quantitative precipitin assay and immunofluorescence.

Biochem/physiol Actions

Albumin is a transport protein that binds a broad range of ligands such as fatty acids, bilirubin, hemin, drugs, amino acids and ions among several others. Albumin may also function as a zinc carrier protein and consequently regulate physiological processes.

Physical form

Supplied as a liquid containing 15mM sodium azide as preservative.

Preparation Note

treated to remove lipoproteins

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A novel mutation in the albumin gene (c.1A>C) resulting in analbuminemia
Caridi G, et al.
European Journal of Clinical Investigation, 43, 72-78 (2013)
Albumin as fatty acid transporter
van der Vusse GJ
Drug Metabolism and Pharmacokinetics, 24, 300-307 (2009)
Maja Thim Larsen et al.
Journal of controlled release : official journal of the Controlled Release Society, 287, 132-141 (2018-07-18)
Recombinant albumin-drug genetic fusions are an effective technology to prolong the serum half-life of therapeutics that has resulted in marketed products. Indirect evidence suggests albumin fusions' long circulation is controlled by engagement with the cellular recycling neonatal Fc receptor (FcRn)
C Magnon et al.
Gene therapy, 15(21), 1436-1445 (2008-06-13)
Given as a prophylactic treatment, a single muscle electrogene transfer of plasmid coding canstatin fused to human serum albumin (CanHSA), slowed down the development of two xenografted human carcinomas from mammary (MDA-MB-231) and prostate origin (PC-3) in nude mice and
N Shaklai et al.
The Journal of biological chemistry, 259(6), 3812-3817 (1984-03-25)
Approximately 10% of the albumin in normal human serum is modified by nonenzymatic glycosylation, primarily at the epsilon-amino group of lysine residue 525. Incubation of albumin with glucose under physiological conditions in vitro resulted in glycosylation of the same residue.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service