P0834
Protein standard
liquid, 2 mg protein/ml
Synonym(s):
Albumin from bovine serum, protein standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
bovine
form
liquid
technique(s)
ELISA: suitable
color
clear colorless
UniProt accession no.
storage temp.
2-8°C
Gene Information
bovine ... ALB(280717)
Looking for similar products? Visit Product Comparison Guide
Application
BSA protein standard has been commonly used in ELISA assays to determine the level of anti-BSA antibodies in serum. Microtiter plates can be coated with 100 μg/ml of BSA protein standard.
Concentrated solution for all protein assays.
Packaging
Sealed ampules provide assurance that the concentration will be correct with every assay.
Physical form
2 mg bovine serum albumin/ml in 0.9% NaCl containing 0.05% sodium azide
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
In vivo (Athens, Greece), 19(2), 375-382 (2005-03-31)
Forty-five approximately 50% in-bred 14-day-old White Leghorn female chickens (Gallus domesticus) originating from 11 hens were distributed into 5 treatment groups containing one sister in each treatment group. Phase I involved oral administration of an antigen, Bovine Serum Albumin (BSA)
Environment international, 158, 106997-106997 (2022-01-08)
Multiple health effects are associated with moisture damage in buildings. Studies explaining these associations and cell-level mechanisms behind the observed health effects are urgently called for. We focused on characterizing gene expression in human airway epithelium after exposure to indoor
Mammalian genome : official journal of the International Mammalian Genome Society (2022-12-25)
Ubiquinol cytochrome c reductase hinge protein (UQCRH) is required for the electron transfer between cytochrome c1 and c of the mitochondrial cytochrome bc1 Complex (CIII). A two-exon deletion in the human UQCRH gene has recently been identified as the cause
International journal of experimental pathology, 95(3), 199-208 (2014-04-26)
Pancreatic acinar cell necrosis is indicative of severe pancreatitis and the degree of necrosis is an index of its outcome. We studied whether the dose and duration of injury correlates with severity, particularly in terms of necrosis, in caerulein-induced acute
Postepy higieny i medycyny doswiadczalnej (Online), 71(0), 952-959 (2017-11-28)
The effect of whey protein hydrolysate (WPH) addition on growth of standard yoghurt cultures and Bifidobacterium adolescentis during co-fermentation and its viability during storage at 4ºC in yoghurts has been evaluated. WPH was obtained with the use of serine protease
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service