Skip to Content
Merck
All Photos(3)

Documents

A7434

Sigma-Aldrich

Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

technique(s)

direct ELISA: 1:40,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:20
western blot (chemiluminescent): 1:30,000 using β-actin in total cell extract of HiLa cells (5-10 μg per well)

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice . Anti-Mouse IgG (Fc specific)-Alkaline Phosphatase antibody is specific for mouse IgG and mouse IgG, Fc fragment. The product does not react with mouse IgG, Fab fragment, human IgG, IgA, and IgM, or rat IgG.

Specificity

Anti-Mouse IgG (Fc specific)-Alkaline Phosphatase antibody is specific for mouse IgG and mouse IgG, Fc fragment. The product does not react with mouse IgG, Fab fragment, human IgG, IgA, and IgM, or rat IgG.

Immunogen

Purified mouse IgG, Fc fragment

Application

Alkaline phosphatase-conjugated goat anti-mouse Fc specific antibody was used as a secondary antibody in ELISA assays at a dilution of 1:1000 in PBS/0.1% Tween and 1% BSA for 1.5 hours at 37°C. Antibody was developed using 4-nitrophenyl phosphate (Sigma) as a substrate for 30 minutes at 37°C.

Other Notes

Antibody adsorbed with human IgG and rat serum proteins.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2, 50% glycerol and 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background staining with human or rat samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dörte Lodka et al.
Annals of the rheumatic diseases, 83(4), 499-507 (2024-01-06)
Anti-neutrophil cytoplasmic autoantibody (ANCA)-associated vasculitides (AAV) are life-threatening systemic autoimmune diseases manifesting in the kidneys as necrotizing crescentic glomerulonephritis (NCGN). ANCA antigens are myeloperoxidase (MPO) or proteinase 3. Current treatments include steroids, cytotoxic drugs and B cell-depleting antibodies. The use
Vidyaramanan Ganesan et al.
Molecular biology of the cell, 30(3), 302-311 (2018-12-06)
Mitochondria exist in an equilibrium between fragmented and fused states that shifts heavily toward fission in response to cellular damage. Nuclear-to-cytoplasmic cyclin C relocalization is essential for dynamin-related protein 1 (Drp1)-dependent mitochondrial fission in response to oxidative stress. This study
A I Bot et al.
Pharmaceutical research, 17(3), 275-283 (2000-05-09)
Delivery of specific antibodies or immunoglobulin constructs to the respiratory tract may be useful for prophylaxis or active treatment of local or systemic disorders. Therefore, we evaluated the utility of lipid-based hollow-porous microparticles (PulmoSpheres) as a potential delivery vehicle for
Jeroen D Langereis et al.
Clinical & translational immunology, 10(4), e1256-e1256 (2021-04-13)
Complete deficiency of alternative pathway (AP) complement factors, explained by homozygous mutations, is a well-known risk factor for invasive bacterial infections; however, this is less obvious for heterozygous mutations. We describe two siblings with a heterozygous NM_001928.3(CFD):c.125C>A p.(Ser42*) mutation in
Pasquale Gallo et al.
International journal of cancer, 113(1), 67-77 (2004-09-24)
The protective efficacy of xenogeneic vaccination with DNA encoding the HER2 oncogene was evaluated in BALB/c mice transgenic for the transforming form of the neu oncogene, which spontaneously develops carcinomas in all mammary glands. Intramuscular injection of either plasmid DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service