Skip to Content
Merck
All Photos(2)

Key Documents

A1418

Sigma-Aldrich

Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgG (Fc specific)–AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

recombinant

expressed in goat

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

should not react with

human

technique(s)

direct ELISA: 1:40,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25
western blot: 1:50,000-1:100,000 using total cell extract of HeLa cells

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibody plays a crucial role in humoral immune responses such as complement activation, phagocytosis, placental transport and cell surface-receptor binding. Anti-mouse IgG (Fc specific)–alkaline phosphatase antibody (diluted 1: 10,000 in PBS-Tween) can be used in ELISA. It is also useful in immunoblotting. Goat anti-mouse IgG (Fc specific)–alkaline phosphatase antibody reacts specifically with mouse IgG Fc fragment, IgG and its subclasses IgG1, IgG2a, IgG2b and IgG3 but not with human IgG, and mouse IgA or IgM.

Immunogen

Purified mouse IgG Fc fragment.

Application

Alkaline phosphatase-conjugated goat anti-mouse Fc specific antibody was used as a secondary antibody in ELISA assays at a dilution of 1:1000 in PBS/0.1% Tween and 1% BSA for 1.5 hours at 37°C. Antibody was developed using 4-nitrophenyl phosphate (Sigma) as a substrate for 30 minutes at 37°C.
Anti-mouse IgG (Fc specific)–alkaline phosphatase antibody can be used in ELISPOT (immunospot) assay. It can also be used in immunohistochemistry and western blot.

Other Notes

Antibody adsorbed with human IgG.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide

Preparation Note

Adsorbed to reduce background with human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Lewis et al.
European journal of immunology, 21(9), 2087-2094 (1991-09-01)
The immune response to cholera toxin B subunit given orally was studied in 13 human volunteers. A serum IgG and IgA antitoxin response was observed, which was boosted by a second immunization. Using an immunospot assay, cells spontaneously secreting anti-toxin
N Miquel et al.
Parasite immunology, 27(3), 79-88 (2005-05-11)
Pigs single inoculated with Ascaris suum eggs expel the majority of larvae between days 14 and 21 post inoculation (p.i.), but the role of the immune system in expulsion is unclear. To investigate the dynamics of immune responses before, during
Ajeet Singh Yadav et al.
Biomicrofluidics, 17(5), 054101-054101 (2023-09-18)
Effective immunotherapies activate natural antitumor immune responses in patients undergoing treatment. The ability to monitor immune activation in response to immunotherapy is critical in measuring treatment efficacy over time and across patient cohorts. Protein arrays are systematically arranged, large collections
V L Motin et al.
Infection and immunity, 62(10), 4192-4201 (1994-10-01)
LcrV (V antigen), a known unstable 37.3-kDa monomeric peptide encoded on the ca. 70-kb Lcr plasmid of Yersinia pestis, Yersinia pseudotuberculosis, and Yersinia enterocolitica, has been implicated as a regulator of the low-calcium response, virulence factor, and protective antigen. In
Marta Baranowska et al.
Vaccine, 33(49), 6988-6996 (2015-09-22)
Vaccination is at present the most efficient way of preventing influenza infections. Currently used inactivated influenza vaccines can induce virus-neutralizing antibodies that are protective against a particular influenza strain, but hamper the induction of cross-protective T-cell responses to later infections.

Articles

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service