Skip to Content
Merck
All Photos(1)

Documents

B3679

Sigma-Aldrich

BCIP®/NBT-Blue

alkaline phosphatase substrate, chromogenic, liquid

Synonym(s):

5-bromo-4-chloro-3-indolyl-phosphate / Nitro blue tetrazolium chloride liquid substrate system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.83

product name

BCIP®/NBT-Purple Liquid Substrate System for Membranes, ready to use alkaline phosphatase substrate

Quality Level

form

liquid

storage temp.

2-8°C

Application

BCIP®/NBT-Purple Liquid Substrate System for Membranes has been used as a substrate for alkaline phosphatase
  • for the detection of secondary antibodies by immunoblot
  • in ELISPOT assay
  • in dot blot analysis
  • in western blot to visualize protein bands on nitrocellulose membrane

Biochem/physiol Actions

5-bromo-4-chloro-3-indolyl phosphate (BCIP)/ nitro blue tetrazolium (NBT) is a chromogenic substrate system for the detection of alkaline phosphatase (AP) enzyme. AP hydrolyses BCIP to form an intermediate leucoindigo compound and is further oxidized by NBT to form 5,5′-dibromo-4,4′-dichloro indigo (BCI). NBT is reduced to an insoluble purple diformazan (DF) and forms NBT-DF. A mixture of NBT-DF and BCI results in the dark purple colored precipitate. AP by hydrolysing many BCIP molecules makes it a highly sensitive method.

Caution

Stable at least one year at 2-8 °C.

Physical form

Ready-to-use.

Legal Information

BCIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xudong Wang et al.
Frontiers in cell and developmental biology, 9, 690502-690502 (2021-08-28)
Melatonin (Mlt) confers potential antitumor effects in various types of cancer. However, to the best of our knowledge, the role of Mlt in the giant cell tumor of bone (GCTB) remains unknown. Moreover, further research is required to assess whether
T W J M Van Herpen et al.
Annals of botany, 102(3), 331-342 (2008-07-16)
Alpha-gliadin proteins are important for the industrial quality of bread wheat flour, but they also contain many epitopes that can trigger celiac (coeliac) disease (CD). The B-genome-encoded alpha-gliadin genes, however, contain very few epitopes. Controlling alpha-gliadin gene expression in wheat
pSTAT5 and ERK exhibit different expression in myeloproliferative neoplasms
Wisniewska-Chudy E, et al.
Oncology Reports, 37(4), 2295-2307 (2017)
Characterization of the 41 kDa allergen Asp v 13, a subtilisin-like serine protease from Aspergillus versicolor
Shi C and Miller JD
Molecular Immunology, 48(15-16), 1827-1834 (2011)
Membrane bioreactor separator system for integrated IgG fragmentation and Fab purification
Yu D and Ghosh R
Journal of Immunological Methods, 359(1-2), 37-41 (2010)

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service