Skip to Content
Merck
All Photos(5)

Documents

A7130

Sigma-Aldrich

β-Amylase from barley

greener alternative

Type II-B, 20-80 units/mg protein (biuret)

Synonym(s):

1,4-α-D-Glucan maltohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type II-B

Quality Level

form

powder

specific activity

20-80 units/mg protein (biuret)

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in starch ethanol research. For more information see the article in biofiles.

Application

β -Amylase is used to hydrolyze α bonds of α-linked polysaccharides, such as starch and glycogen. β -Amylase, has been used in various plant studies, such as carbon starvation studies in Populus tremuloides . β -Amylase, from barley, has been used to study how pressure and temperature affect catalytic activity .

Biochem/physiol Actions

β-Amylase hydrolyzes the α-(1,4) glucan linkages in polysaccharides of three or more α-(1,4) linked D-glucose units. Natural substrates such as starch and glycogen are broken down into glucose and maltose. Pure, crystalline β-amylase preparation consists of four isoenzymes with different isoelectric points. The enzyme polymerizes very rapidly through the sulfhydryl groups in the absence of reducing agents. p-Chloromercuribenzoate inhibits the polymerization and the enzymatic activity. The reducing agents mercaptoethanol or dithiothreitol can completely restore the activity.

Quality

Crude

Unit Definition

One unit will liberate 1.0 mg of maltose from starch in 3 min at pH 4.8 at 20 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volker Heinz et al.
Biotechnology progress, 21(6), 1632-1638 (2005-12-03)
The depolymerization of starch by beta-amylase during exposure to hydrostatic pressure up to 700 MPa and within a temperature range from 20 to 70 degrees C has been investigated. Inactivation of the enzyme as well as alterations in conversion speed
Purification and characterisation of crystalline -amylase from barley.
K Visuri et al.
European journal of biochemistry, 28(4), 555-565 (1972-08-04)
Claudinéia Aparecida Soares et al.
Journal of agricultural and food chemistry, 59(12), 6672-6681 (2011-05-20)
Different banana cultivars were used to investigate the influences of starch granule structure and hydrolases on degradation. The highest degrees of starch degradation were observed in dessert bananas during ripening. Scanning electron microscopy images revealed smooth granule surface in the
Xin Li et al.
Journal of industrial microbiology & biotechnology, 38(11), 1837-1843 (2011-04-21)
A moderately halophilic strain LY9 with high amylolytic activity was isolated from soil sample obtained from Yuncheng, China. Biochemical and physiological characterization along with 16S rRNA sequence analysis placed the isolate in the genus Halobacillus. Amylase production started from the
Heike Reinhold et al.
The Plant cell, 23(4), 1391-1403 (2011-04-14)
Plants contain β-amylase-like proteins (BAMs; enzymes usually associated with starch breakdown) present in the nucleus rather than targeted to the chloroplast. They possess BRASSINAZOLE RESISTANT1 (BZR1)-type DNA binding domains--also found in transcription factors mediating brassinosteroid (BR) responses. The two Arabidopsis

Protocols

Beta-amylase is found in bacteria, fungi, and plants. To measure β-amylase activity, this assay uses a colorimetric spectrophotometric stop reaction at 540 nm.

Beta-amylase is found in bacteria, fungi, and plants. To measure β-amylase activity, this assay uses a colorimetric spectrophotometric stop reaction at 540 nm.

Beta-amylase is found in bacteria, fungi, and plants. To measure β-amylase activity, this assay uses a colorimetric spectrophotometric stop reaction at 540 nm.

Beta-amylase is found in bacteria, fungi, and plants. To measure β-amylase activity, this assay uses a colorimetric spectrophotometric stop reaction at 540 nm.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service