Skip to Content
Merck
All Photos(1)

Key Documents

A4862

Sigma-Aldrich

α-Amylase from Bacillus sp.

liquid

Synonym(s):

1,4-α-D-glucan glucanohydrolase, 1,4-alpha-D-glucan glucanohydrolase, Termamyl® 300L, alpha-amylase, endoglucanase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

Bacillus sp.

Quality Level

form

liquid

storage temp.

2-8°C

General description

α-Amylase belongs to glycosyl hydrolase family 13.[1] It comprises of A, B and C domains. Domain A has two aspartic acid and a glutamic acid as catalytic residues. Domain B possesses Ca2+ and Na+ binding sites.[2]

Application

α-Amylase from Bacillus sp. has been used in the digestion of lyophilized microalgae sample.[3] It has also been used to evaluate α-amylase inhibitory activity in marine algae samples[4] and Myrmecodia platytyrea.[5]

Biochem/physiol Actions

α-Amylase hydrolyzes the polysaccharides starch, malto-oligosaccharides at the α-d-(1,4)-glucosidic linkages to α-anomeric products.[1] It has wide applications in industries including food, pharmaceutical, textile and paper.[6]

Unit Definition

One unit will liberate 1.0 mg of maltose from starch in 3 minutes at pH 6.9 at 20 deg C.

Legal Information

A product of Novozyme Corp.
Termamyl is a registered trademark of Novozymes Corp.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improving the reversibility of thermal denaturation and catalytic efficiency of Bacillus licheniformis alpha-amylase through stabilizing a long loop in domain B
Li Z, et al.
PLoS ONE, 12(3), e0173187-e0173187 (2017)
High-performance thin-layer chromatography-direct bioautography as a method of choice for alpha-amylase and antioxidant activity evaluation in marine algae
Agatonovic-Kustrin S and Morton DW
Journal of Chromatography A, 1530, 197-203 (2017)
Microalgae Characterization for Consolidated and New Application in Human Food, Animal Feed and Nutraceuticals
Molino A, et al.
International Journal of Environmental Research and Public Health, 15(11), 2436-2436 (2018)
High-performance thin-layer chromatographic methods in the evaluation of the antioxidant and anti-hyperglycemic activity of Myrmecodia platytyrea as a promising opportunity in diabetes treatment
Agatonovic-Kustrin S, et al.
Journal of Chromatography A, 1530, 192-196 (2017)
Crystal structure of Bacillus subtilis alpha-amylase in complex with acarbose
Kagawa M, et al.
Journal of Bacteriology, 185(23), 6981-6984 (2003)

Protocols

Follow our procedure for the determination of alpha-Amylase activity. This enzymatic assay of a-Amylase guides you through the entire process and necessary calculations.

Follow our procedure for the determination of alpha-Amylase activity. This enzymatic assay of a-Amylase guides you through the entire process and necessary calculations.

Follow our procedure for the determination of alpha-Amylase activity. This enzymatic assay of a-Amylase guides you through the entire process and necessary calculations.

Follow our procedure for the determination of alpha-Amylase activity. This enzymatic assay of a-Amylase guides you through the entire process and necessary calculations.

Questions

  1. Can I dilute the alpha amylase (A4862) in distilled/deionized water or should I use any specific buffer for dilution?

    1 answer
    1. This product may be diluted further using ultrapure water. Please see the link below to review the enzymatic assay method for this product:
      https://www.sigmaaldrich.com/US/en/technical-documents/protocol/protein-biology/enzyme-activity-assays/enzymatic-assay-of-a-amylase

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service