Skip to Content
Merck
All Photos(1)

Key Documents

A5153

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:7,000-1:21,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Affinity isolated antigen specific antibody is obtained from goat anti-mouse IgG antiserum by immunospecific purification which removes essentially all goat serum proteins, including immunoglobulins, that do not specifically bind to mouse IgG. Goat anti-Mouse IgG is conjugated to Sigma Alkaline Phosphatase, Type VII-S (Product No. P 5521) by protein crosslinking with 0.2% glutaraldehyde. Anti-Mouse IgG (Whole Molecule) is determined to be immunospecific for mouse IgG by immunoelectrophoresis (IEP), versus mouse IgG and normal mouse serum, prior to conjugation. Prior to conjugation, the antibody is found to be reactive with mouse IgG1, IgG2a, IgG2b, and IgG by Ouchterlony Double Diffusion (ODD). Identity and purity of the antibody is established by immunoelectrophoresis (IEP), prior to conjugation. Electrophoresis of the antibody preparation followed by diffusion versus anti-goat IgG and anti-goat whole serum result in single arcs of precipitation.

Specificity

Binds all mouse Igs.
mouse IgG1, IgG2a, IgG2b, and IgG

Immunogen

Purified mouse IgG

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Surfactant Protein A was detected in bronchoalveolar fluid using alkaline phosphatase conjugated goat anti-mouse IgG as the secondary at μg/ml in TBS/Tween containing final concentration of 0.5M NaCl.
Western blot analysis of nuclear or mitochondrial protein extracts were performed using alkaline phosphatase conjugated goat anti-mouse IgG as the secondary antibody at a 1:5000 dilution in 5%BSA/TBS for 1 hour at room temperature. 5-bromo-4-chloro-3-indolyl phosphate/nitroblue tetrazolium (Sigma) was used for the substrate.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tonia Zangari et al.
mBio, 8(2) (2017-03-16)
Epidemiological studies on Streptococcus pneumoniae show that rates of carriage are highest in early childhood and that the major benefit of the pneumococcal conjugate vaccine (PCV) is a reduction in the incidence of nasopharyngeal colonization through decreased transmission within a
Lana J McMillan et al.
Applied and environmental microbiology, 82(2), 538-548 (2015-11-08)
Soluble inorganic pyrophosphatases (PPAs) that hydrolyze inorganic pyrophosphate (PPi) to orthophosphate (Pi) are commonly used to accelerate and detect biosynthetic reactions that generate PPi as a by-product. Current PPAs are inactivated by high salt concentrations and organic solvents, which limits
Siti Aisyah Abdul Ahmad et al.
Virology journal, 14(1), 229-229 (2017-11-23)
The rapid rise and spread in dengue cases, together with the unavailability of safe vaccines and effective antiviral drugs, warrant the need to discover and develop novel anti-dengue treatments. In this study the antiviral activity of geraniin, extracted from the
Dhaval K Shah et al.
International journal of pharmaceutics, 465(1-2), 228-238 (2014-02-11)
To facilitate the development of an inverse targeting strategy, where anti-topotecan antibodies are administered to prevent systemic toxicity following intraperitoneal topotecan, a pharmacokinetic/toxicodynamic (PK/TD) model was developed and evaluated. The pharmacokinetics of 8C2, a monoclonal anti-topotecan antibody, were assessed following
Xian Fu et al.
mBio, 8(5) (2017-09-07)
Methionine sulfoxide reductase A (MsrA) is an antioxidant enzyme found in all domains of life that catalyzes the reduction of methionine-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service