Skip to Content
Merck
All Photos(1)

Key Documents

A3562

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Goat Anti-Mouse IgG (whole molecule)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

goat

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

mouse

technique(s)

direct ELISA: 1:30,000
dot blot: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50
western blot: 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases . Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice . Anti-Mouse IgG (whole molecule)-Alkaline Phosphatase antibody is specific for normal mouse serum and mouse IgG. In Ouchterlony double diffusion assays, the antibody reacts with mouse IgG1, IgG2a, IgG2b, IgG3, IgA, and IgM.

Immunogen

Purified mouse IgG

Application

Anti-Mouse IgG (whole molecule)-Alkaline Phosphatase antibody is suitable for use in direct ELISA (1:3000) and western blot. The product can also be used for immunohistochemistry (1:50 using formalin-fixed, paraffin-embedded sections).
Surfactant Protein A was detected in bronchoalveolar fluid using alkaline phosphatase conjugated goat anti-mouse IgG as the secondary at μg/ml in TBS/Tween containing final concentration of 0.5M NaCl.

Physical form

Solution in 0.05 M Tris buffer, pH 8.0, containing 1 mM MgCl2, 10 mM glycine, 1% bovine serum albumin, 50% glycerol and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Wagh et al.
Cell death & disease, 5, e1320-e1320 (2014-07-11)
FAM40B (STRIP2) is a member of the striatin-interacting phosphatase and kinase (STRIPAK) complex that is involved in the regulation of various processes such as cell proliferation and differentiation. Its role for differentiation processes in embryonic stem cells (ESCs) is till
Jose Velilla et al.
Neurology. Genetics, 5(2), e312-e312 (2019-05-02)
To identify the genetic cause of disease in a form of congenital spinal muscular atrophy and arthrogryposis (CSMAA). A 2-year-old boy was diagnosed with arthrogryposis multiplex congenita, severe skeletal abnormalities, torticollis, vocal cord paralysis, and diminished lower limb movement. Whole-exome
Peng Li et al.
TheScientificWorldJournal, 2014, 174253-174253 (2014-09-10)
This paper describes the complex effects of postmortem ultimate pH (pHu) on Chinese Yellow crossbreed cattle quality during postmortem ageing and provides an explanation of how pHu affects beef tenderness. High pHu beef had the highest initial tenderness (P <
S Munir Alam et al.
Journal of virology, 82(1), 115-125 (2007-10-19)
Two human monoclonal antibodies (MAbs) (2F5 and 4E10) against the human immunodeficiency virus type 1 (HIV-1) envelope g41 cluster II membrane proximal external region (MPER) broadly neutralize HIV-1 primary isolates. However, these antibody specificities are rare, are not induced by
Himanish Ghosh et al.
Nature communications, 7, 13365-13365 (2016-11-08)
GABAA receptors (GABAARs) mediate the majority of fast inhibitory neurotransmission in the brain via synergistic association with the postsynaptic scaffolding protein gephyrin and its interaction partners. However, unlike their counterparts at glutamatergic synapses, gephyrin and its binding partners lack canonical

Articles

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service