Skip to Content
Merck
All Photos(1)

Documents

50020

Sigma-Aldrich

β-Glycerol phosphate disodium salt pentahydrate

≥98.0% (NT)

Synonym(s):

β-GP, β-glycerolphosphate, Disodium β-glycerol phosphate pentahydrate, glycerol-2-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7Na2O6P · 5H2O
CAS Number:
Molecular Weight:
306.11
Beilstein:
3744328
EC Number:
MDL number:
UNSPSC Code:
41141710
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98.0% (NT)

form

solid

impurities

≤2% L-α-glycerol phosphate

loss

26-32% loss on drying

solubility

H2O: 0.1 g/mL, clear, colorless

storage temp.

2-8°C

SMILES string

O.O.O.O.O.[Na+].[Na+].OCC(CO)OP([O-])([O-])=O

InChI

1S/C3H9O6P.2Na.5H2O/c4-1-3(2-5)9-10(6,7)8;;;;;;;/h3-5H,1-2H2,(H2,6,7,8);;;5*1H2/q;2*+1;;;;;/p-2

InChI key

PEMUISUYOHQFQH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

β-Glycerolphosphate (Glycerol-2-Phosphate) is used in the development of hydrogels and scaffolds that have applications in tissue engineering and cell growth and differentiation.

Biochem/physiol Actions

Beta-Glycerophosphate is a classical serine-threonine phosphatase inhibitor used in kinase reaction buffers. BGP is often used in combination with other phosphatase/protease inhibitors for broad spectrum inhibition. It functions as an organic phosphate donor and has been used in culture media for mesenchymal stem cell differentiation to osteoblast-type cells. BGP is also used to buffer M17 media for Lactococcus culture in recombinant protein expression

Other Notes

Substrate for myo-inositol-1-phosphatase from bovine brain

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yung-Hsin Cheng et al.
Tissue engineering. Part A, 16(2), 695-703 (2009-09-23)
Injectable hydrogel is one of the great interests for tissue engineering and cell encapsulation. In the study, the gelatin molecules were added to the thermosensitive chitosan/beta-glycerol phosphate (C/GP) disodium salt hydrogels to form chitosan/gelatin/beta-glycerol phosphate (C/G/GP) disodium salt hydrogels which
A Sanghani-Kerai et al.
Bone & joint research, 6(6), 358-365 (2017-06-04)
Cellular movement and relocalisation are important for many physiologic properties. Local mesenchymal stem cells (MSCs) from injured tissues and circulating MSCs aid in fracture healing. Cytokines and chemokines such as Stromal cell-derived factor 1(SDF-1) and its receptor chemokine receptor type
Yoshitomo Honda et al.
Scientific reports, 3, 3420-3420 (2013-12-07)
Stem cell-based disease modeling presents unique opportunities for mechanistic elucidation and therapeutic targeting. The stable induction of fate-specific differentiation is an essential prerequisite for stem cell-based strategy. Bone morphogenetic protein 2 (BMP-2) initiates receptor-regulated Smad phosphorylation, leading to the osteogenic
Cristiane Machado Mengatto et al.
PloS one, 6(1), e15848-e15848 (2011-01-26)
Successful dental and orthopedic implants require the establishment of an intimate association with bone tissue; however, the mechanistic explanation of how biological systems accomplish osseointegration is still incomplete. We sought to identify critical gene networks involved in osseointegration by exploring
Liesbet Lodewyckx et al.
Arthritis research & therapy, 14(1), R16-R16 (2012-01-24)
The aim of this research was to study molecular changes in the articular cartilage and subchondral bone of the tibial plateau from mice deficient in frizzled-related protein (Frzb) compared to wild-type mice by transcriptome analysis. Gene-expression analysis of the articular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service