Skip to Content
Merck
All Photos(6)

Key Documents

C0875

Sigma-Aldrich

Calcein

Powder

Synonym(s):

Bis[N,N−bis(carboxymethyl)aminomethyl]fluorescein, Fluorescein-bis(methyliminodiacetic acid), Fluorexon

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H26N2O13
CAS Number:
Molecular Weight:
622.53
Beilstein:
8181068
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.

description

mixed isomers

form

powder

technique(s)

titration: suitable

color

yellow to brown

solubility

NaOH: 50 mg/mL

fluorescence

λex 470 nm; λem 509 nm (pH 7.4)

suitability

suitable for (Suitable for fluorometric determination of Ca Suitable for EDTA titration of Ca in the presence of Mg)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OC(=O)CN(CC(O)=O)Cc1cc2c(Oc3cc(O)c(CN(CC(O)=O)CC(O)=O)cc3C24OC(=O)c5ccccc45)cc1O

Looking for similar products? Visit Product Comparison Guide

Application

Calcein has been used:
  • in whole cell staining
  • in histomorphometry and micro-computed tomography
  • to label active bone-forming surfaces

Biochem/physiol Actions

Calcein is a calcium-dependent fluorescent molecule. It is mainly required to study bone metabolism under in vivo conditions and helps in the staining of pit area under in vitro conditions. Calcein is used for the fluorometric determination of calcium and EDTA (ethylenediaminetetraacetic acid) titration of calcium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Parathyroid hormone-related peptide is required for increased trabecular bone volume in parathyroid hormone-null mice.
Miao D, et al.
Endocrinology, 145, 3554-3554 (2004)
Calcium: Chemistry, Analysis, Function and Effects (2015)
Skeletal abnormalities in Pth-null mice are influenced by dietary calcium.
Miao D, et al.
Endocrinology, 145, 2046-2046 (2004)
Wnt10b deficiency results in age-dependent loss of bone mass and progressive reduction of mesenchymal progenitor cells
Stevens JR, et al.
Journal of Bone and Mineral Research, 25(10), 2138-2147 (2010)
South African Topical Ophthalmic Drug Guide (2013)

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service