Skip to Content
Merck
All Photos(1)

Documents

A0878

Sigma-Aldrich

Ala-Gly

≥99% (TLC)

Synonym(s):

L-Alanyl-glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NH2)CONHCH2COOH
CAS Number:
Molecular Weight:
146.14
Beilstein:
1723438
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Ala-Gly,

Assay

≥99% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

C[C@H](N)C(=O)NCC(O)=O

InChI

1S/C5H10N2O3/c1-3(6)5(10)7-2-4(8)9/h3H,2,6H2,1H3,(H,7,10)(H,8,9)/t3-/m0/s1

InChI key

CXISPYVYMQWFLE-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Alanyl dipeptides such as ala-leu, ala-lys, ala-gly, ala-pro, ala-tyr and ala-phe may be used in physicochemical studies or to evaluate dipeptide separation technologies. Alanyl dipeptides may also be used for studying cell uptake mechanisms, dipeptide metabolism or cell growth supplementation benefits.
L-alanylglycine is a simple nutritional dipeptide also used for physical chemistry studies such as hydrogen bonding and heavy metal complexation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masatoshi Watabe et al.
Journal of inorganic biochemistry, 100(10), 1653-1659 (2006-07-22)
We prepared platinum(IV) complexes containing dipeptide and diimine or diamine, the [PtCl(dipeptide-N,N,O)(diimine or diamine)]Cl complex, where -N,N,O means dipeptide coordinated as a tridentate chelate, dipeptide=glycylglycine (NH(2)CH(2)CON(-)CH(2)COO(-), digly, where two protons of dipeptide are detached when the dipeptide coordinates to metal
Analysis of vibrational spectra of L-alanylglycine based on density functional theory calculations.
Padmaja L, Ravikumar C, James C, et al.
Spectrochimica Acta Part A: Molecular Spectroscopy, 71, 252-262 (2008)
M Tafazzoli et al.
Magnetic resonance in chemistry : MRC, 46(4), 370-376 (2008-02-15)
(13)C chemical shieldings and (14)N and (2)H electric field gradient (EFG) tensors of L-alanylglycine (L-alagly) dipeptide were calculated at RHF/6-31 + + G** and B3LYP/6-31 + + G** levels of theory respectively. For these calculations a crystal structure of this
E K Patterson
The Journal of biological chemistry, 264(14), 8004-8011 (1989-05-15)
Bestatin, [(2S,3R)-3-amino-2-hydroxy-4-phenyl-butanoyl]-L-leucine, a known inhibitor of aminopeptidases, is shown to be a potent linear competitive inhibitor (KI,2.7 nM) of a dipeptidase purified from Ehrlich-Lettré hyperdiploid mouse ascites tumor cells. This inhibition can be classified as "slow binding" but not as
W M Zuk et al.
Biopolymers, 28(11), 2025-2044 (1989-11-01)
The CH-stretching vibrational CD (VCD) spectra of glycyl-L-alanine, L-alanylglycine, and L-alanyl-L-alanine have been studied at neutral, high, and low pH in D2O solution. The intense positive VCD band attributed to the C alpha H stretch of the alanyl residue in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service