Skip to Content
Merck
All Photos(1)

Key Documents

73097

Sigma-Aldrich

L-Argininosuccinic acid lithium salt

≥95% (TLC)

Synonym(s):

N-{{[(4S)-4-Amino-4-carboxybutyl]amino}iminomethyl}-L-aspartic acid lithium salt, Lithium L-argininosuccinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18N4O6 · xLi+
Molecular Weight:
290.27 (free acid basis)
Beilstein:
6933810
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥70% (qNMR)
≥95% (TLC)

form

powder or crystals

optical activity

[α]/D 15±2°, c = 1 in H2O

impurities

≤20% water

color

white to off-white

storage temp.

2-8°C

SMILES string

OC(C[C@@H](C(O)=O)NC(NCCC[C@H](N)C(O)=O)=N)=O.[Li+]

InChI

1S/C10H18N4O6.Li/c11-5(8(17)18)2-1-3-13-10(12)14-6(9(19)20)4-7(15)16;/h5-6H,1-4,11H2,(H,15,16)(H,17,18)(H,19,20)(H3,12,13,14);/q;+1/t5-,6-;/m0./s1

InChI key

MGJRREWOOITZBB-GEMLJDPKSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Arginosuccinic acid is a basic amino acid, which is synthesized by some cells from citrulline, aspartic acid and used as a precursor for arginine in the urea cycle or Citrulline-NO cycle. The reaction is catalyzed by the enzyme argininosuccinate synthetase. Argininosuccinic acid is a precursor to fumarate in the citric acid cycle via argininosuccinate lyase. Defects in the arginosuccinate lyase enzyme can lead to arginosuccinate lyase deficiency. Argininosuccinate (ASA) lyase deficiency results in defective cleavage of ASA. This leads to an accumulation of ASA in cells and an excessive excretion of ASA in urine (arginosuccinic aciduria). In virtually all respects, this disorder shares the characteristics of other urea cycle defects. The most important characteristic of ASA lyase deficiency is its propensity to cause hyperammonemia in affected individuals. ASA in affected individuals is excreted by the kidney at a rate practically equivalent to the glomerular filtration rate (GFR). Whether ASA itself causes a degree of toxicity due to hepatocellular accumulation is unknown; such an effect could help explain hyperammonemia development in affected individuals. Regardless, the name of the disease is derived from the rapid clearance of ASA in urine, although elevated levels of ASA can be found in plasma. ASA lyase deficiency is associated with high mortality and morbidity rates.
Metabolite in alanine, aspartate, glutamate, arginine, and proline metabolic pathways, and in the biosynthesis of secondary metabolites and amino acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wing Lok Au et al.
Journal of the neurological sciences, 209(1-2), 101-104 (2003-04-11)
A 25-year-old Chinese man presented with a 2-year history of recurrent coma. His plasma ammonia level was extremely elevated, with raised citrulline level and absence of argininosuccinic acid. Adult-onset citrullinaemia, a condition rarely reported outside the Japanese population, was diagnosed.
L D Fleisher et al.
American journal of human genetics, 31(4), 439-445 (1979-07-01)
We have monitored two successive pregnancies in a family which we found to be at risk for argininosuccinic aciduria. We measured argininosuccinic acid (ASA) concentrations in amniotic fluid and utilized an indirect assay of ASA lyase activity in cultured amniotic
V Reid Sutton et al.
Molecular genetics and metabolism, 78(1), 11-16 (2003-02-01)
Argininosuccinate lyase (AL) has several roles in intermediary metabolism. It is an essential component of the urea cycle, providing a pathway for the disposal of excess nitrogen in mammals. AL links the urea cycle to the tricarboxylic acid (TCA) cycle
W J Kleijer et al.
Journal of inherited metabolic disease, 25(5), 399-410 (2002-11-01)
A biochemical variant of argininosuccinate lyase deficiency, found in five individuals, is introduced. In comparison to classical patients, the variant cases of argininosuccinate lyase deficiency were characterized by residual enzyme activity as measured by the incorporation of [14C]citrulline into proteins.
Rachel S Carling et al.
Clinical chemistry and laboratory medicine, 58(5), 758-768 (2019-10-18)
Background Quantification of plasma amino acids is key to the diagnosis of inherited defects of amino acid synthesis, catabolism and transport, many of which present as clinical emergencies. The utility of this test is limited by the long analysis time

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service