Skip to Content
Merck
All Photos(1)

Key Documents

69453

Sigma-Aldrich

L-Homocysteine

≥98.0% (NT)

Synonym(s):

(S)-2-Amino-4-mercaptobutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
135.18
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

L-Homocysteine, ≥98.0% (NT)

Assay

≥98.0% (NT)

form

powder

optical activity

[α]/D 25.5±1.5°, c = 1 in 1 M HCl

color

white

mp

263-265 °C

application(s)

cell analysis

relevant disease(s)

Alzheimer′s disease

storage temp.

−20°C

SMILES string

N[C@@H](CCS)C(O)=O

InChI

1S/C4H9NO2S/c5-3(1-2-8)4(6)7/h3,8H,1-2,5H2,(H,6,7)/t3-/m0/s1

InChI key

FFFHZYDWPBMWHY-VKHMYHEASA-N

General description

L-homocysteine is used to diagnose homocystinuria.

Application

L-homocysteine has been used as a supplement in cell culture media.

Biochem/physiol Actions

Elevated homocysteine levels in plasma and tissue are associated with brain atrophy in Alzheimer′s disease as well as in healthy eldery subjects. Increased homocysteine is due to deficiencies in vitamin B status, specifically folate, B6 and B12 vitamins, which act as cofactors for enzymes involved in homocysteine metabolism.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Mehlig et al.
Heart (British Cardiac Society), 99(23), 1761-1765 (2013-09-10)
An elevated level of total plasma homocysteine (tHcy) has been associated with risk of coronary heart disease (CHD). The level of tHcy is affected by lifestyle, in addition to genetic predisposition. The methylene tetrahydrofolate reductase (MTHFR) 677C>T polymorphism (rs1801133) is
James J H Chong et al.
Journal of the American Heart Association, 8(1), e011028-e011028 (2019-01-01)
Background Neutrophil gelatinase-associated lipocalin ( NGAL ) or lipocalin 2 may promote atherosclerosis and plaque instability leading to increased risk of cardiac events. We investigated the relationships between plasma NGAL , cardiovascular disease biomarkers, and long-term cardiac events. Methods and
SAGE open medicine, 286054-286054 (2014)
Sanne Visser et al.
American journal of obstetrics and gynecology, 211(4), 373-373 (2014-03-19)
The aim of this study was to investigate which nonclassic cardiovascular biomarkers are associated with persistent endothelial dysfunction after pregnancy in women with a history of hypertensive pregnancy disorders compared with women with uncomplicated pregnancies. This was a systematic review
Rajendra P Shukla et al.
Advanced healthcare materials, 8(15), e1900462-e1900462 (2019-06-27)
The antipsychotic clozapine is the most effective medication available for schizophrenia and it is the only antipsychotic with a known efficacious clinical range. However, it is dramatically underutilized due to the inability to test clozapine blood levels in finger-pricked patients'

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service