Skip to Content
Merck
All Photos(2)

Documents

I2512

Sigma-Aldrich

Sodium iodoacetate

powder, ≥98%

Synonym(s):

Iodoacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICH2COONa
CAS Number:
Molecular Weight:
207.93
Beilstein:
4009322
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

product name

Sodium iodoacetate, ≥98%

biological source

synthetic (organic)

Assay

≥98%

form

powder

mp

208 °C (dec.) (lit.)

solubility

H2O: 100 mg/mL

storage temp.

−20°C

SMILES string

[Na+].[O-]C(=O)CI

InChI

1S/C2H3IO2.Na/c3-1-2(4)5;/h1H2,(H,4,5);/q;+1/p-1

InChI key

AGDSCTQQXMDDCV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the modification of cysteine residues in proteins. Used in protein sequencing (including by Sanger in sequencing bovine insulin) to prevent oxidation of -SH sidechains.
Reagent for the modification of cysteine residues in proteins. Used to induce animal osteoarthritis model system, which mimics both symptoms and histopathology of the human disease.

Biochem/physiol Actions

Used to induce osteoarthritis-like lesions and functional impairment in rats, for testing of chondroprotective agents and diagnostic imaging techniques.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

406.4 °F

Flash Point(C)

208 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Guingamp et al.
Arthritis and rheumatism, 40(9), 1670-1679 (1997-10-27)
To characterize the dose-responsiveness of morphologic and biochemical chondral changes relative to mobility in mono-iodoacetate (MIA)-induced osteoarthritis (OA) in rats. Rat mobility was assessed by biotelemetry. Articular lesions were characterized by macroscopic and histologic examinations. Cartilage proteoglycan metabolism was evaluated
Ikufumi Takahashi et al.
PloS one, 13(4), e0196625-e0196625 (2018-04-27)
This study aimed to investigate the histopathological changes in the patellofemoral joint using a rat model of osteoarthritis that was induced using monosodium iodoacetate, and to establish a novel model of patellofemoral osteoarthritis in a rat model using histopathological analysis.
Matilde Tschon et al.
Cells, 9(7) (2020-07-02)
The purpose of this study was to verify the efficacy of a single intra-articular (i.a.) injection of a hyaluronic acid-chitlac (HY-CTL) enriched with two low dosages of triamcinolone acetonide (TA, 2.0 mg/mL and 4.5 mg/mL), in comparison with HY-CTL alone
S E Bove et al.
Osteoarthritis and cartilage, 11(11), 821-830 (2003-11-12)
To describe an in vivo model in the rat in which change in weight distribution is used as a measure of disease progression and efficacy of acetaminophen and two nonsteroidal anti-inflammatory drugs (NSAIDs) in a model of monosodium iodoacetate (MIA)-induced
Hyung Jin Lim et al.
Food science & nutrition, 8(12), 6550-6556 (2020-12-15)
In Asia, Vigna angularis (azuki bean) has been used as a traditional medicine to treat various diseases because of its biological properties. Osteoarthritis (OA) and osteoporosis (OP) are common regenerative bone diseases that are characterized by deterioration of joint and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service