Skip to Content
Merck
All Photos(1)

Key Documents

60399

Sigma-Aldrich

Potassium iodide

BioUltra, ≥99.5% (AT)

Synonym(s):

KI

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
grade:
reagent grade
form:
solid
solubility:
H2O: 1 M at 20 °C, clear, colorless
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

vapor pressure

1 mmHg ( 745 °C)

product line

BioUltra

Assay

≥99.5% (AT)

form

solid

impurities

insoluble matter, passes filter test
≤0.001% total nitrogen (N)

loss

≤0.5% loss on drying, 110 °C

pH

6.0-9.0 (25 °C, 1 M in H2O)

mp

681 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

bromide, chloride (as Cl-): ≤100 mg/kg
iodate (IO3-): ≤2 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤300 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 273 nm in H2O at 1 M

application(s)

vitamins, nutraceuticals, and natural products

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium iodide has been used:
  • to inhibit active oxygen as a component of processes such as DNA damage[1]
  • as a chaotropic agent which, in high concentrations can dissolve a gel matrix[1]
  • in the recovery of DNA from gel slices by gel dissolution[1]

Biochem/physiol Actions

Potassium iodide (KI) consists of halogen iodine and the alkali metal potassium. It is used in the form of iodized salt to treat endemic goiter. KI is also used to treat hyperthyroidism and radiation protection of the thyroid gland. KI is useful for the preoperative preparation of patients with Grave′s disease. It also finds its use in treating dermatological conditions like cutaneous sporotrichosis and various inflammatory dermatoses. KI is commonly used as an expectorant. It inhibits the capacity of neutrophils to produce e toxic oxygen intermediates hydrogen peroxide and hydroxyl radicals in vitro.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H.O. Smith
Methods in Enzymology, 65, 375-375 (1980)
Isolation of large molecular weight DNA from agarose gels for further digestion by restriction enzymes.
N Blin et al.
FEBS letters, 53(1), 84-86 (1975-04-15)
Higson, S.P.J. et al.
Electroanalysis, 6, 431-431 (1994)
S C Clement et al.
Pediatric blood & cancer, 60(11), 1833-1838 (2013-07-09)
Thyroid dysfunction has been reported in up to 52% of patients 1.4 years after treatment with (131) I-Metaiodobenzylguanidine (MIBG) in children with neuroblastoma (NBL), despite the use of potassium-iodide (KI). Our aim was to investigate if the incidence and severity
J B Sterling et al.
Journal of the American Academy of Dermatology, 43(4), 691-697 (2000-09-27)
Potassium iodide (KI) is a useful drug in the dermatologic armamentarium. It is successfully used for inflammatory dermatoses, most notably erythema nodosum, subacute nodular migratory panniculitis, nodular vasculitis, erythema multiforme, and Sweet's syndrome. KI is also successfully used for cutaneous

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service