Skip to Content
Merck
All Photos(1)

Key Documents

438464

Sigma-Aldrich

Potassium iodate

99.995% trace metals basis

Synonym(s):

Potassium iodate, Potassium iodine oxide (KIO3 )

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KIO3
CAS Number:
Molecular Weight:
214.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.995% trace metals basis
form:
powder and chunks
Pricing and availability is not currently available.

Assay

99.995% trace metals basis

form

powder and chunks

impurities

≤30 ppm Trace metallic impurities

mp

560 °C (lit.)

density

3.93 g/mL at 25 °C (lit.)

SMILES string

[K+].[O-]I(=O)=O

InChI

1S/HIO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

JLKDVMWYMMLWTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Oxidizer used recently to form 2-styrylchromones from o-hydroxy-ω-cinnamylideneacetophenones by oxidative cyclization.[1]

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Singhi, M. Grover, S.K.
Indian J. Chem. B, 33, 1083-1083 (1994)
Mohsen Zeeb et al.
Talanta, 78(2), 584-589 (2009-02-11)
In this work, an on-line system with vapor-phase generation (VPG) and Fourier transform infrared (FTIR) spectrometric detection has been developed as a direct and highly selective analytical technique for the assay of penicillamine (PA). Potassium iodate solution was injected into
C Dryden et al.
Journal of perinatology : official journal of the California Perinatal Association, 27(5), 320-322 (2007-04-25)
Congenital thyrotoxicosis is a rare and potentially fatal illness. We report a case in a preterm infant delivered to a mother known to have autoimmune endocrine disease. Diagnosis was difficult because the infant's presenting symptoms and signs closely resembled congenital
Tatsuya Kawai et al.
Academic radiology, 18(6), 682-689 (2011-03-12)
Evaluation of contrast enhancement of pulmonary lesions with ground-glass attenuation (GGA) is difficult with conventional techniques but might be possible using contrast-mapping images (CMIs) obtained by dual-energy computed tomography. To address this issue, a phantom study was conducted, and this
Qimeng Li et al.
European journal of nutrition, 51(6), 685-691 (2011-09-23)
The aim of the present feeding trial with iodine was to assess pigs' growth performance and carcass characteristics, the iodine accumulation in tissues, and their influences on the thyroid hormones in plasma. Eighty pigs (33-115 kg body weight) were allotted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service