Skip to Content
Merck
All Photos(1)

Key Documents

32319

Sigma-Aldrich

Diethylenetriaminepentaacetic acid

for complexometry, ≥99.0%

Synonym(s):

(Carboxymethylimino)bis(ethylenenitrilo)tetraacetic acid, N,N-Bis(2-[bis(carboxymethyl)amino]ethyl)glycine, DETAPAC, DTPA, Penta(carboxymethyl)diethylenetriamine, Pentetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(HOOCCH2)2NCH2CH2]2NCH2COOH
CAS Number:
Molecular Weight:
393.35
Beilstein:
1810219
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

grade

for complexometry

Assay

≥99.0% (KT)
≥99.0%

ign. residue

≤0.05% (as SO4)

loss

≤0.1% loss on drying, 20°C, HV

mp

219-220 °C (lit.)

anion traces

chloride (Cl-): ≤1000 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤10 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤500 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

suitability

suitable for suitability for complexometry

functional group

amine
carboxylic acid

SMILES string

OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CCN(CC(O)=O)CC(O)=O

InChI

1S/C14H23N3O10/c18-10(19)5-15(1-3-16(6-11(20)21)7-12(22)23)2-4-17(8-13(24)25)9-14(26)27/h1-9H2,(H,18,19)(H,20,21)(H,22,23)(H,24,25)(H,26,27)

InChI key

QPCDCPDFJACHGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Titration Center to learn more.

Application

  • Grafting nanocellulose with diethylenetriaminepentaacetic acid: Development of a novel paper additive for enhancing recycled OCC pulp fibers by functionalizing nanocellulose with DTPA (Li et al., 2022).
  • Controlling multivalency and multimodality: Study on up to pentamodal dendritic platforms based on DTPA cores, illustrating DTPA′s role in multivalency and drug delivery systems (Pulido et al., 2014).
  • Adsorption of Hg (II) ions by DTPA-modified cellulose: Exploration of a DTPA-modified cellulose adsorbent for removing mercury ions from aqueous solutions (Li et al., 2019).
  • DTPA-functionalized multi-walled carbon nanotubes/titanium oxide-PVDF nanofiber membrane: Study on the effective separation of oil/water mixtures using a DTPA-functionalized membrane (Venkatesh et al., 2021).

Other Notes

Complexing agent for the chelation of iron in solutioncitation
Complexing agent for the chelation of iron in solution

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

392.0 °F - closed cup

Flash Point(C)

200 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.R. Bowyer
Febs Letters, 172, 239-239 (1984)
Janice S C Chew-Harris et al.
Annals of clinical biochemistry, 50(Pt 1), 39-46 (2012-11-07)
It is well known that plasma creatinine concentration is affected by muscle mass, while some studies have suggested cystatin C is affected by body mass index (BMI). Our aim was to assess the effects of lean versus fat mass on
Vincent Cutillas et al.
Antimicrobial agents and chemotherapy, 59(1), 310-316 (2014-10-29)
Clinical studies have shown that integrase strand transfer inhibitors (INSTIs) can be used effectively against HIV-1 infection. To date, no resistance substitution has been found in INSTI-naive patients treated with the new integrase inhibitor dolutegravir (DTG). In a recent selection
Yang Song et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(24), 9725-9730 (2009-06-06)
The reactions of glutathione (GSH) with polychlorinated biphenyl (PCB) quinones having different degrees of chlorination on the quinone ring were examined. EPR spectroscopy and MS revealed 2 types of reactions yielding different products: (i) a nonenzymatic, nucleophilic displacement of chlorine
Sébastien Leguay et al.
Inorganic chemistry, 51(23), 12638-12649 (2012-11-17)
Complexation of trivalent actinides with DTPA (diethylenetriamine pentaacetic acid) was studied as a function of pcH and temperature in (Na,H)Cl medium of 0.1 M ionic strength. Formation constants of both complexes AnHDTPA(-) and AnDTPA(2-) (where An stands for Am, Cm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service