Skip to Content
Merck
All Photos(1)

Key Documents

32325

Sigma-Aldrich

Diethylenetriamine-pentaacetic acid calcium trisodium salt hydrate

≥97.0% (dried material, KT)

Synonym(s):

CaNa3-DTPA, Calcium-trisodium diethylenetriamine-pentaacetate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18CaN3Na3O10 · xH2O
CAS Number:
Molecular Weight:
497.35 (anhydrous basis)
Beilstein:
8182304
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (dried material, KT)

form

powder

impurities

~10% water

functional group

amine

SMILES string

[Na+].[Na+].[Na+].[H]O[H].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN1CC(=O)O[Ca]OC(=O)C1

InChI

1S/C14H23N3O10.Ca.3Na.H2O/c18-10(19)5-15(1-3-16(6-11(20)21)7-12(22)23)2-4-17(8-13(24)25)9-14(26)27;;;;;/h1-9H2,(H,18,19)(H,20,21)(H,22,23)(H,24,25)(H,26,27);;;;;1H2/q;+2;3*+1;/p-5

InChI key

AWBWYUNJDQLQGT-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Application

Diethylenetriamine-pentaacetic acid calcium trisodium salt hydrate (CaNa3-DTPA) is a chelator, which can be used as an antidote to metal poisoning. It is generally used to sequester cadmium, beryllium, actinides including plutonium and americium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Competitive binding of Pu and Am with bone mineral and novel chelating agents.
Guilmette R A, et al.
Radiation Protection Dosimetry, 105(1-4), 527-534 (2003)
Chelators as antidotes of metal toxicity: therapeutic and experimental aspects.
Blanusa M, et al.
Current Medicinal Chemistry, 12(23), 2771-2794 (2005)
Influence of methionine administration during chelation of cadmium by CaNa3DTPA and DMPS in the rat1.
Tandon S K, et al.
Environmental Toxicology and Pharmacology, 3(3), 159-165 (1997)
Comparative effectiveness of CaNa 3 DTPA and tiron along with α-tocopherol against beryllium-induced biochemical alterations in rats.
Mathur R, et al.
Indian Journal of Experimental Biology (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service