Skip to Content
Merck
All Photos(4)

Documents

D6518

Sigma-Aldrich

Diethylenetriaminepentaacetic acid

≥99% (titration)

Synonym(s):

(Carboxymethylimino)bis(ethylenenitrilo)tetraacetic acid, N,N-Bis(2-[bis(carboxymethyl)amino]ethyl)glycine, DETAPAC, DTPA, Penta(carboxymethyl)diethylenetriamine, Pentetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(HOOCCH2)2NCH2CH2]2NCH2COOH
CAS Number:
Molecular Weight:
393.35
Beilstein:
1810219
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥99% (titration)

form

powder

mp

219-220 °C (lit.)

solubility

1 M HCl: 50 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CCN(CC(O)=O)CC(O)=O

InChI

1S/C14H23N3O10/c18-10(19)5-15(1-3-16(6-11(20)21)7-12(22)23)2-4-17(8-13(24)25)9-14(26)27/h1-9H2,(H,18,19)(H,20,21)(H,22,23)(H,24,25)(H,26,27)

InChI key

QPCDCPDFJACHGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethylenetriaminepentaacetic acid is a diethylenetriaminepentaacetic anhydride, which is a bifunctional chelating agent. It is used for the treatment of poisoning.

Application

Diethylenetriaminepentaacetic acid has been used:
  • for cofilin activity assay
  • to determine the specificity of superoxide in the transverse aortic constriction (TAC) hearts
  • for Europium bead-based detection assays
  • to detect reduced (S- active) or irreversibly oxidized (SO3H inactive) tyrosine-protein phosphatase non-receptor type 6 (SHP-1) in thymocytes

Preparation Note

Purified

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

392.0 °F - closed cup

Flash Point(C)

200 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection of Intracellular Reduced (Catalytically Active) SHP-1 and Analyses of Catalytically Inactive SHP-1 after Oxidation by Pervanadate or H2O2
Choi S and Love PE
Bio-protocol, 8(1) (2018)
Paraoxonase 2 prevents the development of heart failure
Li W, et al.
Free Radical Biology & Medicine, 121, 117-126 (2018)
Bioconjugate Techniques (2013)
Identification of Novel Protein Targets of Dimethyl Fumarate Modification in Neurons and Astrocytes Reveals Actions Independent of Nrf2 Stabilization
Piroli GG, et al.
Molecular and Cellular Proteomics, mcp-RA118 (2018)
Highly stable lyophilized homogeneous bead-based immunoassays for on-site detection of bio warfare agents from complex matrices
Mechaly A, et al.
Analytical Chemistry, 88(12), 6283-6291 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service