Skip to Content
Merck
All Photos(1)

Key Documents

22640

Sigma-Aldrich

Quinine hemisulfate salt monohydrate

BioReagent, suitable for fluorescence, 99.0-101.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O2 · 0.5H2O4S · H2O
CAS Number:
Molecular Weight:
391.47
Beilstein:
6113937
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

99.0-101.0%

mp

~225 °C (dec.) (lit.)

fluorescence

λex 347 nm; λem 448 nm in 0.5 M sulfuric acid

suitability

suitable for fluorescence

antibiotic activity spectrum

parasites

Mode of action

enzyme | inhibits

SMILES string

O.O.OS(O)(=O)=O.COc1ccc2nccc([C@@H](O)C3CC4CCN3C[C@@H]4C=C)c2c1.COc5ccc6nccc([C@@H](O)C7CC8CCN7C[C@@H]8C=C)c6c5

InChI

1S/2C20H24N2O2.H2O4S.2H2O/c2*1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;1-5(2,3)4;;/h2*3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;(H2,1,2,3,4);2*1H2/t2*13-,14-,19-,20+;;;/m00.../s1

InChI key

ZHNFLHYOFXQIOW-LPYZJUEESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quinine hemisulfate salt monohydrate is a naturally occurring well known fluorescent standard material. Quinine hemisulfate salt monohydrate or QSM has a fluorescence lifetime between 1.0 to 3.0 ns.

Application

Quinine hemisulfate salt monohydrate is used for indirect photometric detection of polyamines in biological samples. It is suitable for studying metabolism of bio crystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.

Biochem/physiol Actions

Potassium channel blocker. Antimalarial
Potassium channel blocker. Antimalarial, anticholinergic, antihypertensive, and hypoglycemic agent; alkaloid originally isolated from the Cinchona family of South American trees. Inhibits mitochondrial ATP-regulated potassium channel. Used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S D Pack et al.
Optics letters, 23(15), 1215-1217 (2007-12-19)
We report on a novel laser-induced fluorescence triple-integration method (LIFTIME) that is capable of making rapid, continuous fluorescence lifetime measurements by a unique photon-counting technique. The LIFTIME has been convolved with picosecond time-resolved laser-induced fluorescence, which employs a high-repetition-rate mode-locked
Y Ma et al.
Journal of chromatography, 608(1-2), 93-96 (1992-09-11)
A rapid separation of polyamines and some related amino acids in cultured tumor cells by high-performance capillary zone electrophoresis with indirect photometric detection is demonstrated. 60 cm x 75 microns I.D. fused-silica capillary was used for the separation and quinine
Kira Behm et al.
Dalton transactions (Cambridge, England : 2003), 46(33), 10867-10875 (2017-08-02)
The synthesis, characterization, and fluorescence spectroscopy of E(C
Elaine B Bohórquez et al.
Malaria journal, 11, 350-350 (2012-10-24)
The naturally fluorescent compound quinine has long been used to treat malaria infections. Although some evidence suggests that quinine acts in the parasite food vacuole, the mechanism of action of quinine has not yet been resolved. The Plasmodium falciparum multidrug
Yuting Bai et al.
Nanoscale research letters, 13(1), 287-287 (2018-09-19)
Carbon dots (CDs) are a member of fluorescent carbon nanomaterials that are widely applied in bioimaging, photothermal therapy (PTT), and biosensors for its tunable fluorescence, photothermal conversion property, and excellent biocompatibility. Surface passivation and doping especially the doping of N

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service