1597005
USP
Quinine sulfate
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Quinine hemisulfate salt monohydrate
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
quinine
mol wt
monograph mol wt. 782.94
manufacturer/tradename
USP
mp
~225 °C (dec.) (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
O.O.OS(O)(=O)=O.COc1ccc2nccc([C@@H](O)C3CC4CCN3C[C@@H]4C=C)c2c1.COc5ccc6nccc([C@@H](O)C7CC8CCN7C[C@@H]8C=C)c6c5
InChI
1S/2C20H24N2O2.H2O4S.2H2O/c2*1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;1-5(2,3)4;;/h2*3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;(H2,1,2,3,4);2*1H2/t2*13-,14-,19-,20+;;;/m00.../s1
InChI key
ZHNFLHYOFXQIOW-LPYZJUEESA-N
Looking for similar products? Visit Product Comparison Guide
General description
Quinine sulfate is the sulfate salt of quinine, an alkaloid, obtained from the bark of Cinchona species.[1][2]
Application
- Quinine Sulfate Capsules
- Quinine Sulfate Tablets
Biochem/physiol Actions
Analysis Note
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service