Skip to Content
Merck
All Photos(1)

Key Documents

12007C

Sigma-Aldrich

Fetal Bovine Serum

Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture

Synonym(s):

Fetal Calf Sera, Fetal Calf Serum, GI FBS, GI FCS, GI Fetal Bovine Sera, GI Fetal Bovine Serum, FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71
biological source:
bovine fetus
origin:
Australia origin
sterility:
γ-irradiated
sterile-filtered
shipped in:
dry ice

biological source

bovine fetus

Quality Level

sterility

γ-irradiated
sterile-filtered

product line

FBS Premier

composition

Hemoglobin, ≤25 mg/dL

origin

Australia origin

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

FBS sourced from Australia is used in a broad range of cell culture applications. FBS provides many non-defined growth promoting and survival enhancing factor to cells in culture. Australia carries the best possible rating in relation to Geographical Bovine Spongiform Encephalopathy (BSE) Risk (GBR) as designated by the Scientific Steering Committee on the Geographical Risk of Bovine Spongiform Encephalopathy. The classification for Australia is GBR Level I.

Preparation Note

Gamma irradiation by SER-TAIN process (25-35kGy).

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Footnote

(Formerly product number F3297)

Legal Information

SER-TAIN is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annelies Jorritsma-Smit et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 143, 105096-105096 (2019-11-02)
Therapeutic vaccination is being explored as a treatment strategy for the treatment of patients with primary or metastatic tumours. We developed a vaccine targeted to Human papillomavirus (HPV)-induced tumours based on recombinant Semliki Forest virus (rSFV) encoding a fusion protein
Robert C Tuckey et al.
The Journal of steroid biochemistry and molecular biology, 112(4-5), 213-219 (2008-11-13)
Cytochrome P450scc (CYP11A1) metabolizes vitamin D3 to 20-hydroxyvitamin D3 as the major product, with subsequent production of dihydroxy and trihydroxy derivatives. The aim of this study was to determine whether cytochrome P450scc could metabolize 1alpha-hydroxyvitamin D3 and whether products were
Perrine Barraud et al.
BMC neuroscience, 9, 24-24 (2008-02-20)
The olfactory epithelium is a neurogenic tissue comprising a population of olfactory receptor neurons that are renewed throughout adulthood by a population of stem and progenitor cells. Because of their relative accessibility compared to intra-cranially located neural stem/progenitor cells, olfactory
Thi A Tran et al.
Journal of neuroinflammation, 5, 14-14 (2008-05-14)
Recent animal and human studies implicate chronic activation of microglia in the progressive loss of CNS neurons. The inflammatory mechanisms that have neurotoxic effects and contribute to neurodegeneration need to be elucidated and specifically targeted without interfering with the neuroprotective
Yoshiyuki Mochida et al.
Matrix biology : journal of the International Society for Matrix Biology, 28(1), 44-52 (2008-12-04)
Decorin (DCN) is one of the major matrix proteoglycans in bone. To investigate the role of DCN in matrix mineralization, the expression of DCN in MC3T3-E1 (MC) cell cultures and the phenotypes of MC-derived clones expressing higher (sense; S-DCN) or

Articles

Find answers to questions (FAQs) about fetal bovine serum (FBS) that is gamma-irradiated by the SER-TAIN™ process. Request a sample of FBS at Sigma-Aldrich.com.

Find answers to questions (FAQs) about fetal bovine serum (FBS) that is gamma-irradiated by the SER-TAIN™ process. Request a sample of FBS at Sigma-Aldrich.com.

Find answers to questions (FAQs) about fetal bovine serum (FBS) that is gamma-irradiated by the SER-TAIN™ process. Request a sample of FBS at Sigma-Aldrich.com.

Find answers to questions (FAQs) about fetal bovine serum (FBS) that is gamma-irradiated by the SER-TAIN™ process. Request a sample of FBS at Sigma-Aldrich.com.

See All

Protocols

Protect your investment with best practices for storing and thawing FBS and other sera. Sample FBS for validation in your cell culture protocol.

Related Content

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service