Skip to Content
Merck
All Photos(1)

Documents

12103C

Sigma-Aldrich

Fetal Bovine Serum

USA origin, suitable for cell culture

Synonym(s):

Fetal Calf Sera, Fetal Calf Serum, FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

bovine fetus

Quality Level

product line

FBS Premier

composition

hemoglobin, ≤20 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

FBS remains a popular media supplement because it provides a wide array of functions in cell culture. FBS delivers nutrients, growth and attachment factors and protects cells from oxidative damage and apoptosis by mechanisms that are difficult to reproduce in serum-free media (SFM) systems.

Application

Fetal Bovine Serum was used in different types of cell culture as a supplement, such as the following:
  • PC12 cells grown in Dulbecco′s modified Eagle′s medium supplemented with Fetal Bovine Serum along with other components.
  • HeLa cells were maintained in Roswell Park Memorial Institute (RPMI) 1640 medium containing Fetal Bovine Serum along with other components.
  • BaF3 Cell Culture (cells were maintained in RPMI 1640 medium supplemented with Fetal Bovine Serum along with other components).
  • Primary hippocampal cell cultures (cells were maintained in Eagle′s minimum essential medium and supplemented with heat-inactivated fetal bovine serum along with other components).
  • HeLa, Cos1 and Cos7 cells were cultured in Dulbecco modified Eagle medium (DMEM) with fetal bovine serum along with other components.
Fetal Bovine Serum (FBS) is the most common serum used to supplement cell culture media due to its high nutritional content. Although it is relatively low in protein, FBS is effective in promoting and sustaining growth of vertebrate mammalian and insect cells.

This product is for further manufacturing use. THIS PRODUCT IS NOT INTENDED FOR HUMAN OR THERAPEUTIC USE.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Julie L Webb et al.
The Journal of biological chemistry, 278(27), 25009-25013 (2003-04-30)
Parkinson's disease (PD) is characterized by the loss of dopaminergic neurons in the substantia nigra and the formation of aggregates (Lewy bodies) in neurons. alpha-Synuclein is the major protein in Lewy bodies and rare mutations in alpha-synuclein cause early-onset PD.
Joseph E Rabinowitz et al.
Journal of virology, 76(2), 791-801 (2001-12-26)
The serotypes of adeno-associated virus (AAV) have the potential to become important resources for clinical gene therapy. In an effort to compare the role of serotype-specific virion shells on vector transduction, we cloned each of the serotype capsid coding domains
R J Mark et al.
Journal of neurochemistry, 68(1), 255-264 (1997-01-01)
Peroxidation of membrane lipids results in release of the aldehyde 4-hydroxynonenal (HNE), which is known to conjugate to specific amino acids of proteins and may alter their function. Because accumulating data indicate that free radicals mediate injury and death of
Tadepally Lakshmikanth et al.
Cell reports, 32(3), 107923-107923 (2020-07-23)
The human immune system varies extensively between individuals, but variation within individuals over time has not been well characterized. Systems-level analyses allow for simultaneous quantification of many interacting immune system components and the inference of global regulatory principles. Here, we
D B Bregman et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(21), 11586-11590 (1996-10-15)
Damage to actively transcribed DNA is preferentially repaired by the transcription-coupled repair (TCR) system. TCR requires RNA polymerase II (Pol II), but the mechanism by which repair enzymes preferentially recognize and repair DNA lesions on Pol II-transcribed genes is incompletely

Protocols

Protect your investment with best practices for storing and thawing FBS and other sera. Sample FBS for validation in your cell culture protocol.

Protect your investment with best practices for storing and thawing FBS and other sera. Sample FBS for validation in your cell culture protocol.

Protect your investment with best practices for storing and thawing FBS and other sera. Sample FBS for validation in your cell culture protocol.

Protect your investment with best practices for storing and thawing FBS and other sera. Sample FBS for validation in your cell culture protocol.

Related Content

Our broad range of the most trusted tools for cell culture includes stringently sourced and tested FBS, established media formulations, and sterile labware. Cutting-edge techniques using stem cells and 3D matrices are enabled by organoids, hydrogels, culture scaffolds, and bioinks for 3D bioprinting.

Our broad range of the most trusted tools for cell culture includes stringently sourced and tested FBS, established media formulations, and sterile labware. Cutting-edge techniques using stem cells and 3D matrices are enabled by organoids, hydrogels, culture scaffolds, and bioinks for 3D bioprinting.

Our broad range of the most trusted tools for cell culture includes stringently sourced and tested FBS, established media formulations, and sterile labware. Cutting-edge techniques using stem cells and 3D matrices are enabled by organoids, hydrogels, culture scaffolds, and bioinks for 3D bioprinting.

Our broad range of the most trusted tools for cell culture includes stringently sourced and tested FBS, established media formulations, and sterile labware. Cutting-edge techniques using stem cells and 3D matrices are enabled by organoids, hydrogels, culture scaffolds, and bioinks for 3D bioprinting.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service