Skip to Content
Merck
All Photos(1)

Key Documents

F6765

Sigma-Aldrich

Fetal Bovine Serum

USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture

Synonym(s):

Fetal Calf Serum, FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71
biological source:
bovine fetus
origin:
USA origin
sterility:
sterile-filtered
shipped in:
dry ice
Pricing and availability is not currently available.

biological source

bovine fetus

Quality Level

sterility

sterile-filtered

product line

FBS Select

composition

Bovine IgG, ≤1 mg/mL
Hemoglobin, ≤20 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin (typically ≤1 EU/ml)

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

FBS sourced in the United States is used in a broad range of cell culture applications. FBS provides many non-defined growth promoting and survival enhancing factor to cells in culture. Charcoal stripped FBS is often used instead of FCS to minimize the level of androgen and other hormones provided in 10% serum supplemented media.

Biochem/physiol Actions

Charcoal Stripped Fetal Bovine Serum (FBS) is available for researchers requiring decreased levels of various hormones in their studies. Dextran treated Charcoal is used to selectively remove hormones without nonspecific loss of other serum components. The effectiveness of hormone removal is assessed on the basis of reductions in the levels of T3, T4, and Progesterone pre and post treatment.

This product is designed for researchers involved in studying processes influenced by steroid hormones such as estrogen stimulation and the obesity process and other areas that benefit from reduced hormone levels such as certain types of viral infections.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Other Notes

The product is not available in Australia. Please view our protocol for Charcoal-stripping FBS to deplete hormones here.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andras Franko et al.
Genes, 11(10) (2020-10-11)
Prostate cancer (PCa), the most incident cancer in men, is tightly regulated by endocrine signals. A number of different PCa cell lines are commonly used for in vitro experiments, but these are of diverse origin, and have very different cell-proliferation
Cyril Corbet et al.
Nature communications, 11(1), 454-454 (2020-01-25)
Acidosis, a common characteristic of the tumor microenvironment, is associated with alterations in metabolic preferences of cancer cells and progression of the disease. Here we identify the TGF-β2 isoform at the interface between these observations. We document that acidic pH promotes
Jonathan T Lei et al.
Cell reports, 24(6), 1434-1444 (2018-08-09)
RNA sequencing (RNA-seq) detects estrogen receptor alpha gene (ESR1) fusion transcripts in estrogen receptor-positive (ER+) breast cancer, but their role in disease pathogenesis remains unclear. We examined multiple ESR1 fusions and found that two, both identified in advanced endocrine treatment-resistant
Christoph Lammel et al.
Biomolecules, 10(9) (2020-08-23)
Twenty natural remedies traditionally used against different inflammatory diseases were probed for their potential to suppress the expression of the inflammatory markers E-selectin and VCAM-1 in a model system of IL-1 stimulated human umbilical vein endothelial cells (HUVEC). One third
Ana M Reyes-Ramos et al.
Cancers, 11(10) (2019-10-30)
The paracrine interaction between tumor cells and adjacent stroma has been associated with the oncogenic activity of the Hedgehog (Hh) pathway in triple-negative breast tumors. The present study developed a model of paracrine Hh signaling and examined the impact of

Articles

Review the basics of using fetal bovine (or calf) serum for cell culture and how sourcing and regulations affect the price of sera for cell-based assays.

Protocols

Protect your investment with best practices for storing and thawing FBS and other sera. Sample FBS for validation in your cell culture protocol.

Related Content

Our broad range of the most trusted tools for cell culture includes stringently sourced and tested FBS, established media formulations, and sterile labware. Cutting-edge techniques using stem cells and 3D matrices are enabled by organoids, hydrogels, culture scaffolds, and bioinks for 3D bioprinting.

Our broad range of the most trusted tools for cell culture includes stringently sourced and tested FBS, established media formulations, and sterile labware. Cutting-edge techniques using stem cells and 3D matrices are enabled by organoids, hydrogels, culture scaffolds, and bioinks for 3D bioprinting.

Our broad range of the most trusted tools for cell culture includes stringently sourced and tested FBS, established media formulations, and sterile labware. Cutting-edge techniques using stem cells and 3D matrices are enabled by organoids, hydrogels, culture scaffolds, and bioinks for 3D bioprinting.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

See All

Questions

1–2 of 2 Questions  
  1. Once a bottle of Fetal Bovine Serum (FBS) is thawed, for how long can the bottle be used?

    1 answer
    1. Once Fetal Bovine Serum has been thawed, the bottle can be used in cell culture for up to 7 days for most cell types when stored  refrigerated at 2-8°C.  If the cells are not fastidous (e.g. HeLa or fibroblasts), the thawed and refrigerated serum can be used for up to 30 days.  For some extremely fastidious cell lines, it may be necessary to screen individual lots for acceptable performance.  For uses such as blocking for various immuno procedures, usage will generally be for 30 days and perhaps somewhat longer when stored at 2-8°C.  For other procedures, usage beyond 30 days storage at 2-8°C will need to be determined.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service