Skip to Content
Merck
All Photos(4)

Documents

E7023

Sigma-Aldrich

Ethyl alcohol, Pure

greener alternative

≥99.45%, for molecular biology, 200 proof, suitable for RNA extraction and DNA extraction

Synonym(s):

Ethanol, absolute alcohol, non-denatured ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
EC Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
NACRES:
NA.21

product name

Ethyl alcohol, Pure, 200 proof, for molecular biology

grade

for molecular biology

Quality Level

vapor density

1.59 (vs air)

Assay

≥99.45%

form

liquid

expl. lim.

3.1-27.7 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

200 proof

technique(s)

DNA extraction: suitable
RNA extraction: suitable

impurities

≤0.2% (water)

pH

7.0 (20 °C, 10 g/L)

bp

78.3 °C

mp

-114 °C

density

0.789 g/mL at 20 °C

application(s)

agriculture

greener alternative category

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is 200proof, non-denatured absolute ethanol (100%). This is a molecular biology grade product and is tested for the RNase and Dnase.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Ethanol is environmentally preferable green solvent and thus it aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". 

Application

Pure ethyl alcohol (ethanol) has been used in the following process:
  • DNA and RNA extraction
  • array hybridization
  • ethanol sedation assay
Suitable for use in the precipitation of nucleic acids

Biochem/physiol Actions

Positive allosteric modulator of GABAA receptors, and negative allosteric modulator of NMDA glutamate receptors.

Other Notes

Sales restrictions may apply

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaishree Garhyan et al.
BMC research notes, 6, 72-72 (2013-03-01)
Hybridization based assays and capture systems depend on the specificity of hybridization between a probe and its intended target. A common guideline in the construction of DNA microarrays, for instance, is that avoiding complementary stretches of more than 15 nucleic
Derek J Stefanik et al.
Nature protocols, 8(5), 892-899 (2013-04-13)
Among marine invertebrates, the starlet sea anemone Nematostella vectensis has emerged as an important laboratory model system. One advantage of working with this species relative to many other marine invertebrates is the ease of isolating relatively pure DNA, RNA and
Ross B Mounsey et al.
Experimental neurology, 273, 36-44 (2015-08-06)
Parkinson's disease (PD) is a common chronic neurodegenerative disorder, usually of idiopathic origin. Symptoms including tremor, bradykinesia, rigidity and postural instability are caused by the progressive loss of dopaminergic neurons in the nigrostriatal region of the brain. Symptomatic therapies are
Cem Kuscu et al.
Nature biotechnology, 32(7), 677-683 (2014-05-20)
RNA-guided genome editing with the CRISPR-Cas9 system has great potential for basic and clinical research, but the determinants of targeting specificity and the extent of off-target cleavage remain insufficiently understood. Using chromatin immunoprecipitation and high-throughput sequencing (ChIP-seq), we mapped genome-wide
N Di Girolamo et al.
Stem cells (Dayton, Ohio), 33(1), 157-169 (2014-06-27)
Stem cell (SC) division, deployment, and differentiation are processes that contribute to corneal epithelial renewal. Until now studying the destiny of these cells in a living mammal has not been possible. However, the advent of inducible multicolor genetic tagging and

Protocols

Whole genome amplification (WGA) of plasma and serum DNA presents a unique challenge due to the small amount of nucleic acid in such samples.

Whole genome amplification (WGA) of plasma and serum DNA presents a unique challenge due to the small amount of nucleic acid in such samples.

Whole genome amplification (WGA) of plasma and serum DNA presents a unique challenge due to the small amount of nucleic acid in such samples.

Whole genome amplification (WGA) of plasma and serum DNA presents a unique challenge due to the small amount of nucleic acid in such samples.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service