Skip to Content
Merck
All Photos(5)

Documents

459844

Sigma-Aldrich

Ethyl alcohol, Pure

greener alternative

≥99.5%, ACS reagent, 200 proof

Synonym(s):

Ethanol, absolute alcohol, non-denatured ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
EC Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
NACRES:
NA.21

product name

Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%

grade

ACS reagent

Quality Level

vapor density

1.59 (vs air)

Assay

≥99.5%

form

liquid

expl. lim.

3.1-27.7 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

200 proof

impurities

H2SO4, passes test (darkened)
MnO4- reducers, passes test
acetone, passes test
isopropanol, passes test
≤0.0002 meq/g Titr. base
≤0.0005 meq/g Titr. acid
≤0.1% Methanol
≤0.2% water

evapn. residue

≤0.001%

color

APHA: ≤10

pH

7.0 (20 °C, 10 g/L)

bp

78.3 °C

mp

-114 °C

solubility

H2O: passes test

density

0.789 g/mL at 20 °C

application(s)

agriculture

greener alternative category

storage temp.

room temp

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chromatin isolation by RNA purification (ChIRP). Ci C Chu, Jeffrey J Quinn, Howard Y HY Chang, Journal of Visualized Experiments 2012-01-01
Chromatin isolation by RNA purification (ChIRP)

Genomic maps of long noncoding RNA occupancy reveal principles of RNA-chromatin interactions.
Ci C Chu, Kun K Qu, Franklin L FL Zhong, Steven E SE Artandi, Howard Y HY Chang, Molecular Cell 2011-11-18Genomic maps of long noncoding RNA occupancy reveal principles of RNA-chromatin interactions.
Prices include Federal Tax.
Pure ethyl alcohol is a non-denatured 200proof ethanol. Ethanol, a primary alcohol, is an important raw material for the synthesis of various compounds. It can be synthesized either from molasses by fermentation or by hydration of ethylene. It acts as a hydrogen source and reaction medium during the transfer hydrogenation of carbonyl compounds in the presence of sodium hydroxide. The impact of ethanol/water fumigation on the performance of diesel engines has been studied. ACS grade solvent can be used for processes requiring strict quality conditions such as analytical testing.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Ethanol is environmentally preferable green solvent and thus it aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". 

Application

Pure ethyl alcohol (Ethanol) has been used in the following processes:
  • Preparation of AMBF3 (ammoniomethyltrifluoroborate)-conjugated biomolecules.
  • As an absorbing liquid for the determination of Grüneisen parameter by photoacoustic measurement in an integrating sphere system.
  • Solid-phase microextraction (SPME) on-fiber derivatization analysis of resveratrol extracted from red wine.

Other Notes

Sales restrictions may apply

replacement part

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Photoacoustic measurement of the Gruneisen parameter using an integrating sphere.
Villanueva Y, et al.
The Review of Scientific Instruments, 85(7), 074904-074904 (2014)
Assessment of a direct injection diesel engine fumigated with ethanol/water mixtures.
Morsy MH.
Energy Conversion and Management , 94, 406-414 (2015)
Sodium hydroxide-catalyzed transfer hydrogenation of carbonyl compounds and nitroarenes using ethanol or isopropanol as both solvent and hydrogen donor.
Wang D, et al.
J. Mol. Catal. A: Chem., 400, 14-21 (2015)
Zhibo Liu et al.
Nature protocols, 10(9), 1423-1432 (2015-08-28)
Herein we present a general protocol for the functionalization of biomolecules with an organotrifluoroborate moiety so that they can be radiolabeled with aqueous (18)F fluoride ((18)F(-)) and used for positron emission tomography (PET) imaging. Among the β(+)-emitting radionuclides, fluorine-18 ((18)F)
Ross B Mounsey et al.
Experimental neurology, 273, 36-44 (2015-08-06)
Parkinson's disease (PD) is a common chronic neurodegenerative disorder, usually of idiopathic origin. Symptoms including tremor, bradykinesia, rigidity and postural instability are caused by the progressive loss of dopaminergic neurons in the nigrostriatal region of the brain. Symptomatic therapies are

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service