Skip to Content
Merck
All Photos(7)

Key Documents

793566

Sigma-Aldrich

Sodium chloride

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

Agency

suitable for GB 5009.253-2016

vapor pressure

1.33 hPa ( 865 °C)

product line

ReagentPlus®
Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

pH

7

mp

801 °C (lit.)

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium chloride can be used as:
  • A reagent in the solvent-free chlorination of arenes to synthesize chlorinated arenes in the presence of (diacetoxyiodo)benzene as an oxidant.
  • A catalyst for the one-pot multicomponent reaction of aromatic aldehydes, malononitrile, and thiophenol to synthesize substituted pyridines.
  • A source of electrophilic chlorine to synthesize chlorinated thiophenes, selenophenes, and benzo[b]selenophenes via copper-mediated chlorocyclization.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vandana Tripathy et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 54(5), 394-406 (2019-02-14)
Extraction and quantification of pesticide residue from the milk matrix at or below the established maximum residue limit (MRL) is a challenging task for both analytical chemists and the regulatory institutions to take corrective actions for the human health and
Andreia de Araújo Morandim-Giannetti et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 106(4), 1558-1566 (2017-08-02)
Hydrogels are made from natural or synthetic polymers and, currently, they have many biomedical applications. In this work, the conditions for obtaining a hydrogel with similar physicochemical characteristics to the vitreous humor were defined using polyvinyl alcohol (PVA) and glutaraldehyde
Stefanie Kruse et al.
Journal of chromatography. A, 1640, 461929-461929 (2021-02-21)
Imaging high-performance thin-layer chromatography (HPTLC) was explored with regard to its ability to visualize changes in the metabolite profile of bacteria. Bacillus subtilis has become a model organism in many fields. The increasing interest in these bacteria is driven by
Arnau Vidal et al.
Toxins, 12(2) (2020-02-28)
Deoxynivalenol is one of the most ubiquitous mycotoxins in the Western diet through its presence in cereals and cereal products. A vast amount of studies indicate the worrying level of exposure to this toxin, while even high percentages of the

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service