Skip to Content
Merck
All Photos(5)

Key Documents

S9888

Sigma-Aldrich

Sodium chloride

ACS reagent, ≥99.0%

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder

grade

ACS reagent

Agency

suitable for EPA 1613
suitable for EPA 300
suitable for GB 5009.253-2016

vapor pressure

1.33 hPa ( 865 °C)

Assay

≥99.0%

form

powder

impurities

≤0.005% Insoluble matter

pH

5.0-9.0 (25 °C, 5% in solution)

mp

801 °C (lit.)

anion traces

bromide (Br-): ≤0.01%
chlorate, nitrate (as NO3-): ≤0.003%
iodide (I-): ≤0.002%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.004%

cation traces

Ba:, passes test
Ca: ≤0.002%
Fe: ≤2 ppm
K: ≤0.005%
Mg: ≤0.001%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Sodium chloride has been used for the preparation of tris buffered saline, phosphate buffered saline, MPM-2 (mitotic protein monoclonal 2) cell lysis buffer, immunoprecipitation wash buffer, LB (Luria-Bertani) media and dialysis buffer.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher S Hale et al.
The Review of scientific instruments, 90(3), 034102-034102 (2019-04-01)
The membrane concentration osmometer coupled with multiple sample preparations has been used for over a century to determine a number of colloidal properties. At the dilute region, this method has been used to determine solute molecular mass. When the solution
Immunofluorescent Staining of Mouse Intestinal Stem Cells.
O'Rourke KP et al.
Bio-protocol, 6, e1732-e1732 (2016)
Heterologous expression and purification of catalytic domain of CESA1 from Arabidopsis thaliana.
Vandavasi VG and O?Neill H
Bio-protocol null
MPM-2 Mediated Immunoprecipitation of Proteins Undergoing Proline-directed Phosphorylation.
Antonelli R and Zacchi P
Bio-protocol null
Meng Yuan et al.
Cell host & microbe, 25(6), 873-883 (2019-06-14)
The fusion peptide (FP) of HIV-1 envelope glycoprotein (Env) is essential for mediating viral entry. Detection of broadly neutralizing antibodies (bnAbs) that interact with the FP has revealed it as a site of vulnerability. We delineate X-ray and cryo-electron microscopy

Articles

HPLC-UV method detects 9 antioxidants like propyl gallate in food using a Discovery®

HPLC-UV method detects 9 antioxidants like propyl gallate in food using a Discovery®

HPLC-UV method detects 9 antioxidants like propyl gallate in food using a Discovery®

HPLC-UV method detects 9 antioxidants like propyl gallate in food using a Discovery®

Protocols

Protocol for the preparation of Antibody Sensitized Sheep Erythrocytes. Includes product numbers and links.

Protocol for the preparation of Antibody Sensitized Sheep Erythrocytes. Includes product numbers and links.

Protocol for the preparation of Antibody Sensitized Sheep Erythrocytes. Includes product numbers and links.

Protocol for the preparation of Antibody Sensitized Sheep Erythrocytes. Includes product numbers and links.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service