Skip to Content
Merck
All Photos(4)

Key Documents

S5886

Sigma-Aldrich

Sodium chloride

BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
grade:
reagent
form:
powder or crystals
solubility:
water: soluble (358 g/l at 20 °C (68 °F))

grade

reagent

Quality Level

product line

BioReagent

Assay

≥99%

form

powder or crystals

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable
cell culture | plant: suitable

pH

7

mp

801 °C (lit.)

solubility

water: soluble (358 g/l at 20 °C (68 °F))

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

This product is cell culture tested and is appropriate for use in cell culture applications.

Sodium chloride (NaCl) has been used:
  • for the preparation of phosphate buffer saline
  • in the preservative solution prepared for diluting semen

Other Notes

Easily compare specifications for Sodium Chloride products with the Sodium Chloride specification table.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oil red-O stains non-adipogenic cells: a precautionary note.
Kinkel AD et al.
Cytotechnology, 46, 49-49 (2004)
Quality control of laboratory methods for semen evaluation in a multicenter research study.
Brazil C et al.
Journal of Andrology, 25, 645-645 (2004)
Lan Lin et al.
PLoS pathogens, 15(6), e1007795-e1007795 (2019-06-07)
Infection with the Streptococcus suis (S. suis) epidemic strain can cause Streptococcal toxic shock-like syndrome (STSLS), which is characterized by a cytokine storm, dysfunction of multiple organs and a high incidence of mortality despite adequate treatment. Despite some progress concerning
Frédéric Lebrun-Julien et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(25), 8432-8448 (2014-06-21)
The mammalian target of rapamycin (mTOR) pathway integrates multiple signals and regulates crucial cell functions via the molecular complexes mTORC1 and mTORC2. These complexes are functionally dependent on their raptor (mTORC1) or rictor (mTORC2) subunits. mTOR has been associated with
Alexander W Wyatt et al.
Genome biology, 15(8), 426-426 (2014-08-27)
Genomic analyses of hundreds of prostate tumors have defined a diverse landscape of mutations and genome rearrangements, but the transcriptomic effect of this complexity is less well understood, particularly at the individual tumor level. We selected a cohort of 25

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service