Skip to Content
Merck
All Photos(5)

Key Documents

230391

Sigma-Aldrich

Magnesium sulfate heptahydrate

ACS reagent, ≥98%

Synonym(s):

Epsom salts

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgSO4 · 7H2O
CAS Number:
Molecular Weight:
246.47
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38030409
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98%
98.0-102.0% (ACS specification)
grade:
ACS reagent
form:
crystals

grade

ACS reagent

Quality Level

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥98%
98.0-102.0% (ACS specification)

form

crystals

impurities

≤0.005% insolubles

pH

5.0-8.2 (25 °C, 5%)

anion traces

chloride (Cl-): ≤5 ppm
nitrate (NO3-): ≤0.002%

cation traces

Ca: ≤0.02%
Fe: ≤5 ppm
K: ≤0.005%
Mn: ≤5 ppm
NH4+: ≤0.002%
Na: ≤0.005%
Sr: ≤0.005%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

WRUGWIBCXHJTDG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium sulfate heptahydrate is a hydrated salt of magnesium. It is commonly used as a catalyst in organic synthesis.

Application

Magnesium sulfate heptahydrate can be used as a Lewis acid catalyst to synthesize:      
  • Phenazine and quinoxaline derivatives via a condensation reaction between o-phenylenediamines and 1,2-dicarbonyl compounds.
  • Imidazole derivatives via one-pot multicomponent condensation reaction of aromatic aldehydes, benzil, aliphatic, and aromatic primary amines.

NaBH4 supported on MgSO4 7H2O can be used as an efficient catalyst for the reduction of α, β-unsaturated carbonyl compounds to corresponding alcohols.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Experimental investigation of the stability and evaporation of sulfate and chloride brines on Mars.
Altheide T, et al.
Earth and Planetary Science Letters, 282(1), 69-78 (2009)
Primer Extension Analysis of HBV DNA with Strand-Specific Primers.
Jung J and Kim K.
Bio-protocol, 5(15) (2015)
ESR studies of (Cu(H2O)6)2+ in magnesium sulfate heptahydrate.
Radhakrishna S and Bhaskar Rao T.
Journal of Magnetic Resonance, 32(1), 71-81 (1978)
Benjamin S Waldman et al.
Cell, 180(2), 359-372 (2020-01-21)
Toxoplasma gondii chronically infects a quarter of the world's population, and its recrudescence can cause life-threatening disease in immunocompromised individuals and recurrent ocular lesions in the immunocompetent. Acute-stage tachyzoites differentiate into chronic-stage bradyzoites, which form intracellular cysts resistant to immune
Sulfates on Mars: A systematic Raman spectroscopic study of hydration states of magnesium sulfates.
Wang A, et al.
Geochimica et Cosmochimica Acta, 70(24), 6118-6135 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service